Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VLHIRTIPG HEFLLQSDHE TELRAWHRAL RTVIERLVRW VEARREAPTG RDQGSGDREN PLELRLSGSG PAELSAGEDE EEESELVSKP LLRLSSRRSS IRGPEGTEQN RVRNKLKRLI AKRPPLQSLQ ERGLLRDQVF GCQLESLCQR EGDTVPSFLR LCIAAVDKRG LDVDGIYRVS GNLAVVQKLR FLVDRERAVT SDGRYVFPEQ PGQEGRLDLD STEWDDIHVV TGALKLFLRE LPQPLVPPLL LPHFRAALAL SESEQCLSQI QELIGSMPKP NHDTLRYLLE HLCRVIAHSD KNRMTPHNLG IVFGPTLFRP EQETSDPAAH ALYPGQLVQL MLTNFTSLFP (402-750 aa encoded by BC006107)
This gene encodes a member of the Rho-GAP family of GTPase activating proteins. The protein has substantial GAP activity towards several Rho-family GTPases in vitro, converting them to an inactive GDP-bound state. It is implicated in regulating adhesion of hematopoietic cells to the extracellular matrix. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Rho GTPase-activating protein 9; Rho-type GTPase-activating protein 9
Gene Aliases: 10C; ARHGAP9; RGL1
UniProt ID: (Human) Q9BRR9
Entrez Gene ID: (Human) 64333
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support