Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The antibody detects endogenous levels of total ARHGEF19 protein.
Guanine nucleotide exchange factors (GEFs) such as ARHGEF19 accelerate the GTPase activity of Rho GTPases (see RHOA, MIM 165390).[supplied by OMIM, Dec 2008].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ephexin-2; Rho guanine nucleotide exchange factor (GEF) 19; Rho guanine nucleotide exchange factor 19; weakly similar to Rho GEF
Gene Aliases: ARHGEF19; WGEF
UniProt ID: (Human) Q8IW93
Entrez Gene ID: (Human) 128272
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support