Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: QTEADCINNI NDFLKGCATL QVEIFDPDDL YSGVNFSKVL STLLAVNKAT EDQLSERPCG RSSSLSAANT SQTNPQGAVS STVSGLQRQS KTVEMTENGS HQLIVKARFN FKQTN
Acts as a RAC1 guanine nucleotide exchange factor (GEF).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-Pix; COOL-2; PAK-interacting exchange factor alpha; PAK-interacting exchange factor, alpha; Rac/Cdc42 guanine exchange factor (GEF) 6; Rac/Cdc42 guanine nucleotide exchange factor (GEF) 6; Rac/Cdc42 guanine nucleotide exchange factor 6; Rho guanine nucleotide exchange factor 6
Gene Aliases: alpha-PIX; alphaPIX; ARHGEF6; Cool-2; COOL2; KIAA0006; MRX46; PIXA
UniProt ID: (Human) Q15052
Entrez Gene ID: (Human) 9459
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support