Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: TSKYPSRDMY RESENSSFPS HRHQEKLHVN YLTSLHLQDK KSAAAEAPTD DQPTDLSLPK NPHKPTGKVL GLAHSTTGPQ ESKGISQFQV LGSQSRDCHP KACRVSPMTM SGPKKYPESL SRSGKPHHVR LENFRKME
Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 24%.
ARID5B encodes a member of the AT-rich interaction domain (ARID) family of DNA binding proteins. The encoded protein forms a histone H3K9Me2 demethylase complex with PHD finger protein 2 and regulates the transcription of target genes involved in adipogenesis and liver development. ARID5B also plays a role in cell growth and differentiation of B-lymphocyte progenitors, and single nucleotide polymorphisms in this gene are associated with acute lymphoblastic leukemia. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ARID domain-containing protein 5B; AT rich interactive domain 5B (MRF1-like); AT-rich interactive domain 5B (MRF1-like); AT-rich interactive domain-containing protein 5B; Modulator recognition factor 2; modulator recognition factor 2 (MRF2); MRF-2; MRF1-like protein
Gene Aliases: ARID5B; DESRT; MRF-2; MRF2
UniProt ID: (Human) Q14865
Entrez Gene ID: (Human) 84159
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support