Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Rat
ARL6IP4 is a protein coding gene involved in modulating alternative pre-mRNA splicing with either 5' distal site activation or preferential use of 3' proximal site. In case of infection by Herpes simplex virus (HSVI), may act as a splicing inhibitor of HSVI pre-mRNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADP-ribosylation factor GTPase 6 interacting protein 4; ADP-ribosylation factor like GTPase 6 interacting protein 4; ADP-ribosylation factor-like 6 interacting protein 4; ADP-ribosylation factor-like protein 6-interacting protein 4; ADP-ribosylation-like factor 6 interacting protein 4; aip-4; ARL-6-interacting protein 4; HSP-975; HSVI binding protein; HSVI-binding protein; Splicing factor SRrp37; splicing factor, arginine/serine-rich 20; splicing regulator SRrp38; SR-15; SR-25; SRp25; SRp25 nuclear protein
Gene Aliases: ARL6IP4; SFRS20; SR-25; SRp25; SRrp37
UniProt ID: (Human) Q66PJ3
Entrez Gene ID: (Human) 51329
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support