Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • ARL8A/ARL8B Antibodies

          Proteintech

          ARL8A/ARL8B Monoclonal Antibody (2F4C3)

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite ARL8A/ARL8B Monoclonal Antibody (2F4C3)

          • Antibody Testing Data (3)
          ARL8A/ARL8B Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          ARL8A/ARL8B Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ARL8A/ARL8B Antibody (68186-1-IG) in WB

          Various lysates were subjected to SDS PAGE followed by western blot with 68186-1-Ig (ARL8A/ARL8B antibody) at dilution of 1:20000 incubated at room temperature for 1.5 hours. The membrane was stripped and reblotted with HRP-conjugated GAPDH Monoclonal antibody (HRP-60004) as loading control. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ARL8A/ARL8B Antibody in Western Blot (WB)
          ARL8A/ARL8B Antibody in Western Blot (WB)
          ARL8A/ARL8B Antibody in Western Blot (WB)

          Product Details

          68186-1-IG

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:5,000-1:50,000
          -

          ELISA (ELISA)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rabbit, Rat

          Host/Isotype

          Mouse / IgG2a

          Class

          Monoclonal

          Type

          Antibody

          Clone

          2F4C3

          Immunogen

          Recombinant protein
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Purification

          Protein A

          Storage buffer

          PBS with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice

          Product Specific Information

          Immunogen sequence: MIALFNKLLD WFKALFWKEE MELTLVGLQY SGKTTFVNVI ASGQFNEDMI PTVGFNMRKI TKGNVTIKLW DIGGQPRFRS MWERYCRGVS AIVYMVDAAD QEKIEASKNE LHNLLDKPQL QGIPVLVLGN KRDLPGALDE KELIEKMNLS AIQDREICCY SISCKEKDNI DITLQWLIQH SKSRRS

          Target Information

          ARL8A and ARL8B are members of the ADP-ribosylation factor-like (ARL) family of small GTPases, which play important roles in intracellular trafficking and vesicular transport. The structure of ARL8A and ARL8B consists of a conserved GTPase domain that binds to guanine nucleotides (GDP and GTP) and a C-terminal lipid modification site that allows their association with cellular membranes. They share a high degree of sequence similarity, with ARL8B being the closest homolog of ARL8A. Functionally, ARL8A and ARL8B are involved in the regulation of lysosomal positioning and trafficking. They are primarily localized to late endosomes and lysosomes, where they interact with various effector proteins to control the movement of these organelles along microtubules. ARL8A and ARL8B have been implicated in mediating lysosomal fusion events, lysosomal tubulation, and lysosome-related processes such as autophagy.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ADP-ribosylation factor like GTPase 8A; ADP-ribosylation factor like GTPase 8B; ADP-ribosylation factor-like 10B; ADP-ribosylation factor-like 10C; ADP-ribosylation factor-like 8A; ADP-ribosylation factor-like 8B; ADP-ribosylation factor-like protein 10B; ADP-ribosylation factor-like protein 10C; ADP-ribosylation factor-like protein 8A; ADP-ribosylation factor-like protein 8B; Novel small G protein indispensable for equal chromosome segregation 1; Novel small G protein indispensable for equal chromosome segregation 2; small G protein indispensable for equal chromosome segregation 1; small G protein indispensable for equal chromosome segregation 2

          View more View less

          Gene Aliases: 1110033P22Rik; 2610313E07Rik; 3100002J04Rik; ARL10B; ARL10C; ARL8A; ARL8B; GIE1; GIE2; RGD1565940

          View more View less

          UniProt ID: (Human) Q96BM9, (Human) Q9NVJ2, (Mouse) Q8VEH3, (Rat) Q66HA6, (Mouse) Q3TXD7

          View more View less

          Entrez Gene ID: (Human) 127829, (Human) 55207, (Mouse) 68724, (Rat) 498234, (Rat) 500282, (Mouse) 67166

          View more View less

          Function(s)
          GTPase activity protein binding GTP binding alpha-tubulin binding beta-tubulin binding GDP binding nucleotide binding
          Process(es)
          cell cycle chromosome segregation small GTPase mediated signal transduction lysosome localization cell division mitotic nuclear division biological_process
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7fd48b9cb8-69xq6:80/100.66.134.130:80.
          git-commit: fe0b5dd8e7a67b57f27a6e2d26c18f8954d91d59
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0