Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MIALFNKLL DWFKALFWKE EMELTLVGLQ YSGKTTFVNV IASGQFNEDM IPTVGFNMRK ITKGNVTIKL WDIGGQPRFR SMWERYCRGV SAIVYMVDAA DQEKIEASKN ELHNLLDKPQ LQGIPVLVLG NKRDLPGALD EKELIEKMNL SAIQDREICC YSISCKEKDN IDITLQWLIQ HSKSRRS (1-186 aa encoded by BC015408)
Plays a role in lysosome motility. In neurons, mediates the anterograde axonal long-range transport of presynaptic lysosome-related vesicles required for presynaptic biogenesis and synaptic function. May play a role in chromosome segregation. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADP-ribosylation factor like GTPase 8A; ADP-ribosylation factor-like 10B; ADP-ribosylation factor-like 8A; ADP-ribosylation factor-like protein 10B; ADP-ribosylation factor-like protein 8A; Novel small G protein indispensable for equal chromosome segregation 2; small G protein indispensable for equal chromosome segregation 2
Gene Aliases: 1110033P22Rik; ARL10B; ARL8A; GIE2
UniProt ID: (Human) Q96BM9, (Mouse) Q8VEH3
Entrez Gene ID: (Human) 127829, (Mouse) 68724
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support