Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Aliquoting is unnecessary for -20°C storage.
ARL8B may play a role in lysosome motility. may play a role in chromosome segregation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADP-ribosylation factor like GTPase 8B; ADP-ribosylation factor-like 10C; ADP-ribosylation factor-like 8B; ADP-ribosylation factor-like protein 10C; ADP-ribosylation factor-like protein 8B; Novel small G protein indispensable for equal chromosome segregation 1; small G protein indispensable for equal chromosome segregation 1
Gene Aliases: 2610313E07Rik; 3100002J04Rik; ARL10C; ARL8B; GIE1
UniProt ID: (Human) Q9NVJ2, (Rat) Q66HA6, (Mouse) Q3TXD7
Entrez Gene ID: (Human) 55207, (Pig) 100515580, (Rat) 500282, (Mouse) 67166
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support