Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: AETLAYLIEP DVELQRIASI TDHLIAMLAD YFKYPSSVSA ITDIKRLDHD LKHAHELRQA AFKLYASLGA NDEDIRKKVS LGEGRPPVLT ASRQGVTST
ARMC8 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Armadillo repeat-containing protein 8; GID complex subunit 5, VID28 homolog
Gene Aliases: ARMC8; GID5; HSPC056; S863-2; VID28
UniProt ID: (Human) Q8IUR7
Entrez Gene ID: (Human) 25852
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support