Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ARRDC2 gene ontology (GO) annotation includes plasma membrane and plasma membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Arrestin domain-containing protein 2; induced by lysergic acid diethylamide-1
Gene Aliases: 4632416I05Rik; ARRDC2; CLONE24945; ILAD1; PP2703
UniProt ID: (Human) Q8TBH0, (Mouse) Q9D668
Entrez Gene ID: (Human) 27106, (Mouse) 70807, (Rat) 306344
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support