Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ARRDC1, ARRDC2 (which exists as multiple alternatively spliced isoforms), ARRDC4 and ARRDC5 are arrestin domain-containing proteins that are encoded by genes which map to human chromosomes 9, 15 and 19. Chromosome 9, on which the ARRDC1 gene is localized, contains 145 million base pairs and comprises 4% of the human genome, encoding nearly 900 genes. Hereditary hemorrhagic telangiectasia, which is characterized by harmful vascular defects, and Familial dysautonomia, are both associated with chromosome 9.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Arrestin domain-containing protein 4; liver regeneration-related protein LRRG041/LRRGT00117
Gene Aliases: 2410003C09Rik; Ab1-209; ARRDC4; AV216361; LRRG00117
UniProt ID: (Human) Q8NCT1, (Mouse) A0A0B4J1F4
Entrez Gene ID: (Human) 91947, (Rat) 293019, (Mouse) 66412
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support