Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: MGNGGRSGLQ QGKGNVDGVA ATPTAASASC QYRCIECNQE AKELYRDYNH GVLKITICKS CQKPVDKYIE YDPVIILINA ILCKAQAYRH ILFNTQINIH GKLCIFCLLC EAYLRWWQLQ DSNQNTAPDD LIRYAKEWDF YRMFAIAALE QTAYFIGIFT FLWVERPMTA KKKPNFILLL KALLLSSYGK LLLIPAVIWE HDYTSVCLKL IKVFVLTSNF QAIRVTLNIN RKLSFLAVLS GLLLESIMVY FFQSMEWDVG SDYAIFKSQD F
May act as a mediator of sterol homeostasis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ARV1 fatty acid homeostatsis modulator; ARV1 homolog, fatty acid homeostatsis modulator; hARV1; Protein ARV1
Gene Aliases: ARV1; HT035; RGD1310264
UniProt ID: (Human) Q9H2C2
Entrez Gene ID: (Human) 64801, (Rat) 292097
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support