Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: LSPGGFDDST LPLVDKSLEG EKTGSRDVIP MDALGPDGYS TVDRRERRPR GASSAGEASE KEPLKLDPSR KAPPPGPSRP AVRLVDAVGD AKPQPVDSWV
Involved in protein-protein interactions at adherens junctions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Armadillo repeat protein deleted in velo-cardio-facial syndrome; Splicing regulator ARVCF
Gene Aliases: ARVCF
UniProt ID: (Human) O00192
Entrez Gene ID: (Human) 421
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support