Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: VTSVRPAAQP EICYQLLLNH GAARIYPPQF HKVIQACHSC PKAIEVVVNA YEHIRWNTKW RRAIPDDDLE KYWDFYHSLF TVCCNSPRTL MHLSRCAIRR TLHNRCHRA
Probable substrate-recognition component of a SCF-like ECS.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ankyrin repeat and SOCS box protein 4; ankyrin repeat and SOCS box-containing 4; ASB-4
Gene Aliases: ASB-4; ASB4
UniProt ID: (Human) Q9Y574
Entrez Gene ID: (Human) 51666
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support