Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: LLYAGADVQK GKYWDTPLHA AAQQSSTEIV NLLLEFGADI NAKNTELLRP IDVATSSSMV ERILLQHEAT PSSLYQLCRL CIRSYIGKPR LHLIPQLQLP TLLKNFLQY
ASB5 encodes a protein that is a member of the ankyrin repeat and SOCS box-containing (ASB) family of proteins. They contain ankyrin repeat sequence and SOCS box domain. The SOCS box serves to couple suppressor of cytokine signaling (SOCS) proteins and their binding partners with the elongin B and C complex, possibly targeting them for degradation. Multiple alternatively spliced transcript variants have been described for this gene but their full length sequences are not known.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ankyrin repeat and SOCS box protein 5; ankyrin repeat and SOCS box-containing 5; ankyrin repeat and SOCs box-containing protein 5; ASB-5; SOCS box protein ASB-5
Gene Aliases: 1110018D09Rik; ASB-5; ASB5
UniProt ID: (Human) Q8WWX0, (Mouse) Q9D1A4
Entrez Gene ID: (Human) 140458, (Mouse) 76294
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support