Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VLAMAAAAA PPPVKDCEIE PCKKRKKDDD TSTCKTITKY LSPLGKTRDR VFAPPKPSNI LDYFRKTSPT NEKTQLGKEC KIKSPESVPV DSNKDCTTPL EMFSNVEFKK KRKRVNLSHQ LNNIKTENEA PIGISSDDSK EDYSLNNDFV ESSTSVLRYK KQVEVLAENI QDTKSQPNTM TSLQNSKKVN PKQGTTKNDF KKLRKRKCRD VVDLSESLPL AEELNLLKKD GKDTKQMENT TSHANSRDNV TEAAQLNDSI ITVSYEEFLK SHKENKVEEI PDSTMSICVP SETVDEIVKS GYISESENSE ISQQVRFKTV TVLAQVH (4-329 aa encoded by BC015051)
Involved in DNA damage response. Involved in a RAD9A-related damage checkpoint, a pathway that is important in determining whether DNA damage is compatible with cell survival or whether it requires cell elimination by apoptosis. Modulates the RAD9A interaction with BCL2 and thereby induces DNA damages-induced apoptosis. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATPase family AAA domain-containing protein 5; chromosome fragility associated gene 1; Chromosome fragility-associated gene 1 protein; enhanced level of genomic instability 1 homolog; FLJ12735
Gene Aliases: ATAD5; C17orf41; ELG1; FRAG1
UniProt ID: (Human) Q96QE3
Entrez Gene ID: (Human) 79915
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support