Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MEDSHKSTT SETAPQPGSA VQGAHISHIA QQVSSLSESE ESQDSSDSIG SSQKAHGILA RRPSYRKILK DLSSEDTRGR KGDGENSGVS AAVTSMSVPT PIYQTSSGQY IAIAPNGALQ LASPGTDGVQ GLQTLTMTNS GSTQQGTTIL QYAQTSDGQQ ILVPSNQVVV QTASGDMQTY QIRTTPSATS LPQTVVMTSP VTLTSQTTKT DDPQLKREIR LMKNREAARE CRRKKKEYVK CLENRVAVLE NQNKTLIEEL KTLKDLYSNK SV (1-271 aa encoded by BC029619)
This gene encodes an activating transcription factor, which belongs to the ATF subfamily and bZIP (basic-region leucine zipper) family. It influences cellular physiologic processes by regulating the expression of downstream target genes, which are related to growth, survival, and other cellular activities. This protein is phosphorylated at serine 63 in its kinase-inducible domain by serine/threonine kinases, cAMP-dependent protein kinase A, calmodulin-dependent protein kinase I/II, mitogen- and stress-activated protein kinase and cyclin-dependent kinase 3 (cdk-3). Its phosphorylation enhances its transactivation and transcriptional activities, and enhances cell transformation. Fusion of this gene and FUS on chromosome 16 or EWSR1 on chromosome 22 induced by translocation generates chimeric proteins in angiomatoid fibrous histiocytoma and clear cell sarcoma. This gene has a pseudogene on chromosome 6.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Activating transcription factor 1; cAMP-dependent transcription factor ATF-1; Cyclic AMP-dependent transcription factor ATF-1; Protein TREB36
Gene Aliases: ATF1; EWS-ATF1; FUS/ATF-1; TREB36
UniProt ID: (Human) P18846
Entrez Gene ID: (Human) 466
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support