Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Component of the RNA polymerase II elongator complex, a multiprotein complex associated with the RNA polymerase II (Pol II) holoenzyme, and which is involved in transcriptional elongation (PubMed:22854966). The elongator complex catalyzes formation of carboxymethyluridine in the wobble base at position 34 in tRNAs (PubMed:29332244). Involved in cell migration. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Angiotonin-transactivated protein 1; Elongator complex protein 6; elongator complex protein 6 homolog; Protein TMEM103; transmembrane protein 103; UPF0405 protein C3orf75; UPF0405 protein C3orf75 homolog
Gene Aliases: 2610001P13Rik; 2610002I17Rik; ATP1; AV159994; C3orf75; ELP6; RGD1305478; TMEM103
UniProt ID: (Human) Q0PNE2, (Mouse) Q8BK75, (Rat) B2RYG8
Entrez Gene ID: (Human) 54859, (Mouse) 72341, (Rat) 363150
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support