Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • ATP10B Antibodies

          Abnova

          ATP10B Polyclonal Antibody, MaxPab™

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite ATP10B Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (1)
          ATP10B Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          ATP10B Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ATP10B Antibody (H00023120-B01P) in WB

          Western Blot analysis of ATP10B expression in transfected 293T cell line by ATP10B MaxPab polyclonal antibody. Lane 1: ATP10B transfected lysate(160.8 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ATP10B Antibody in Western Blot (WB)

          Product Details

          H00023120-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          ATP10B (AAI67815.1, 1 a.a. approximately 1461 a.a) full-length human protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MALSVDSSWH RWQWRVRDGF PHCPSETTPL LSPEKGRQSY NLTQQRVVFP NNSIFHQDWE EVSRRYPGNR TCTTKYTLFT FLPRNLFEQF HRWANLYFLF LVILNWMPSM EVFHREITML PLAIVLFVIM IKDGMEDFKR HRFDKAINCS NIRIYERKEQ TYVQKCWKDV RVGDFIQMKC NEIVPADILL LFSSDPNGIC HLETASLDGE TNLKQRCVVK GFSQQEVQFE PELFHNTIVC EKPNNHLNKF KGYMEHPDQT RTGFGCESLL LRGCTIRNTE MAVGIVIYAG HETKAMLNNS GPRYKRSKIE RRMNIDIFFC IGILILMCLI GAVGHSIWNG TFEEHPPFDV PDANGSFLPS ALGGFYMFLT MIILLQVLIP ISLYVSIELV KLGQVFFLSN DLDLYDEETD LSIQCRALNI AEDLGQIQYI FSDKTGTLTE NKMVFRRCTI MGSEYSHQEN AKRLETPKEL DSDGEEWTQY QCLSFSARWA QDPATMRSQK GAQPLRRSQS ARVPIQGHYR QRSMGHRESS QPPVAFSSSI EKDVTPDKNL LTKVRDAALW LETLSDSRPA KASLSTTSSI ADFFLALTIC NSVMVSTTTE PRQRVTIKPS SKALGTSLEK IQQLFQKLKL LSLSQSFSST APSDTDLGES LGANVATTDS DERDDASVCS GGDSTDDGGY RSSMWDQGDI LESGSGTSLE EALEAPATDL ARPEFCYEAE SPDEAALVHA AHAYSFTLVS RTPEQVTVRL PQGTCLTFSL LCTLGFDSVR KRMSVVVRHP LTGEIVVYTK GADSVIMDLL EDPACVPDIN MEKKLRKIRA RTQKHLDLYA RDGLRTLCIA KKVVSEEDFR RWASFRREAE ASLDNRDELL METAQHLENQ LTLLGATGIE DRLQEGVPDT IATLREAGIQ LWVLTGDKQE TAVNIAHSCR LLNQTDTVYT INTENQETCE SILNCALEEL KQFRELQKPD RKLFGFRLPS KTPSITSEAV VPEAGLVIDG KTLNAIFQGK LEKKFLELTQ YCRSVLCCRS TPLQKSMIVK LVRDKLRVMT LSIGDGANDV SMIQAADIGI GISGQEGMQA VMSSDFAITR FKHLKKLLLV HGHWCYSRLA RMVVYYLYKN VCYVNLLFWY QFFCGFSSST MIDYWQMIFF NLFFTSLPPL VFGVLDKDIS AETLLALPEL YKSGQNSECY NLSTFWISMV DAFYQSLICF FIPYLAYKGS DIDVFTFGTP INTISLTTIL LHQAMEMKTW TIFHGVVLLG SFLMYFLVSL LYNATCVICN SPTNPYWVME GQLSNPTFYL VCFLTPVVAL LPRYFFLSLQ GTCGKSLISK AQKIDKLPPD KRNLEIQSWR SRQRPAPVPE VARPTHHPVS SITGQDFSAS TPKSSNPPKR KHVEESVLHE QRCGTECMRD DSCSGDSSAQ LSSGEHLLGP NRIMAYSRGQ TDMCRCSKRS SHRRSQSSLT I

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ATPase class V type 10B; ATPase, class V, type 10B; P4-ATPase flippase complex alpha subunit ATP10B; Phospholipid-transporting ATPase VB

          View more View less

          Gene Aliases: ATP10B; ATPVB; KIAA0715

          View more View less

          UniProt ID: (Human) O94823

          View more View less

          Entrez Gene ID: (Human) 23120

          View more View less

          Function(s)
          magnesium ion binding phospholipid-translocating ATPase activity protein binding ATP binding
          Process(es)
          phospholipid translocation
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0