Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAWRCPRMGR VPLAWCLALC GWACMAPRGT QAEESPFVGN PGNITGARGL TGTLRCQLQV QGEPPEVHWL RDGQILELAD STQTQVPLGE DEQDDWIVVS QLRITSLQLS DTGQYQCLVF LGHQTFVSQP GYVGLEGLPY FLEEPEDRTV AANTPFNLSC QAQGPPEPVD LLWLQDAVPL ATAPGHGPQR SLHVPGLNKT SSFSCEAHNA KGVTTSRTAT ITVLPQQPRN LHLVSRQPTE LEVAWTPGLS GIYPLTHCTL QAVLSDDGMG IQAGEPDPPE EPLTSQASVP PHQLRLGSLH PHTPYHIRVA CTSSQGPSSW THWLPVETPE GVPLGPPENI SATRNGSQAF VHWQEPRAPL QGTLLGYRLA YQGQDTPEVL MDIGLRQEVT LELQGDGSVS NLTVCVAAYT AAGDGPWSLP VPLEAWRPGQ AQPVHQLVKE PSTPAFSWPW WYVLLGAVVA AACVLILALF LVHRRKKETR YGEVFEPTVE RGELVVRYRV RKSYSRRTTE ATLNSLGISE ELKEKLRDVM VDRHKVALGK TLGEGEFGAV MEGQLNQDDS ILKVAVKTMK IAICTRSELE DFLSEAVCMK EFDHPNVMRL IGVCFQGSER ESFPAPVVIL PFMKHGDLHS FLLYSRLGDQ PVYLPTQMLV KFMADIASGM EYLSTKRFIH RDLAARNCML NENMSVCVAD FGLSKKIYNG DYYRQGRIAK MPVKWIAIES LADRVYTSKS DVWSFGVTMW EIATRGQTPY PGVENSEIYD YLRRGNRLKQ PADCLDGLYA LMSRCWELNP QDRPSFTELR EDLENTLKAL PPAQEPDEIL YVNMDEGGGY PEPPGAAGGA DPPTQPDPKD SCSCLTAAEV HPAGRYVLCP STTPSPAQPA DRGSPAAPGQ EDGA
AXL is a chronic myelogenous leukemia-associated oncogene that is also associated with colon cancer and melanoma. The AXL protein is a receptor tyrosine kinase that transduces signals from the extracellular matrix into the cytoplasm by binding growth factors like vitamin K-dependent protein growth-arrest-specific gene 6, and is involved in the stimulation of cell proliferation. TAM receptors are also involved in phagocytosis of apoptotic cells. Mice lacking all three TAM receptors have several degenerative phenotypes linked to inefficient removal of apoptotic cells and membranes (e.g., in the retina and the male reproductive tract) and develop a severe autoimmune phenotype akin to systemic lupus erythematosus, including the production of broad spectrum auto-antibodies. In addition, Axl may function as a putative entry receptor for filoviruses. Axl is also used to identify a specific subpopulation of human blood dendritic cells, also referred to as Siglec 6+/Axl+ dendritic cells. Cellular expression of Axl can be upregulated by TLR ligands, such as LPS or poly I:C. Soluble Axl is generated by proteolytic cleavage of the membrane form. Increased plasma levels may be indicative of inflammation and cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AXL oncogene; AXL transforming sequence/gene; AZF1; oncogene AXL; sAXL; soluble AXL; Tyrosine-protein kinase receptor UFO
Gene Aliases: ARK; AXL; JTK11; Tyro7; UFO
UniProt ID: (Human) P30530
Entrez Gene ID: (Human) 558
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support