Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
OriGene
ASCL3 is a protein coding gene, transcriptional repressor, and inhibits myogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: achaete-scute complex homolog 3; Achaete-scute homolog 3; ASH-3; bHLH transcription factor Sgn-1 (Salivary Glands 1); bHLH transcriptional regulator Sgn-1; bHLHa42; Class A basic helix-loop-helix protein 42
Gene Aliases: ASCL3; BHLHA42; HASH3; SGN1
UniProt ID: (Human) Q9NQ33
Entrez Gene ID: (Human) 56676
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support