Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Actin Muscle Antibodies

          NeoBiotechnologies

          Actin, Muscle Specific (Muscle Cell Marker) Monoclonal Antibody (HHF35)

          View all (18) Actin Muscle antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Actin, Muscle Specific (Muscle Cell Marker) Monoclonal Antibody (HHF35)

          • Antibody Testing Data (5)
          Actin, Muscle Specific (Muscle Cell Marker) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Actin, Muscle Specific (Muscle Cell Marker) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 5

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Actin, Muscle Specific (Muscle Cell Marker) Antibody (MSM1-445-P1ABX) in IHC (P)

          Formalin-fixed, paraffin-embedded human Rhabdomyosarcoma stained with Muscle Specific Actin Mouse Monoclonal Antibody (HHF35). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Actin, Muscle Specific (Muscle Cell Marker) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Actin, Muscle Specific (Muscle Cell Marker) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Actin, Muscle Specific (Muscle Cell Marker) Antibody in Western Blot (WB)
          Actin, Muscle Specific (Muscle Cell Marker) Antibody in Western Blot (WB)
          Actin, Muscle Specific (Muscle Cell Marker) Antibody in SDS-PAGE (SDS-PAGE)

          Product Details

          MSM1-445-P1ABX

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-dependent
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          Assay-dependent
          -

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          0.25-0.5 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          1-2 µg/mL
          -

          Flow Cytometry (Flow)

          1-2 µg/10^6 cells
          -
          Product Specifications

          Species Reactivity

          Dog, Chicken, Cat, Human, Mouse, Rabbit, Rat

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          HHF35

          Immunogen

          SDS extract of human myocardium.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A/G

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          The product encoded by this gene belongs to the actin family of proteins, which are highly conserved proteins that play a role in cell motility, structure and integrity. Alpha, beta and gamma actin isoforms have been identified, with alpha actins being a major constituent of the contractile apparatus, while beta and gamma actins are involved in the regulation of cell motility. This actin is an alpha actin that is found in skeletal muscle. Mutations in this gene cause nemaline myopathy type 3, congenital myopathy with excess of thin myofilaments, congenital myopathy with cores, and congenital myopathy with fiber-type disproportion, diseases that lead to muscle fiber defects.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: a-actin; actin alpha 1; actin alpha cardiac; actin alpha cardiac 1; actin, alpha 1, skeletal muscle; Actin, alpha cardiac muscle 1; Actin, alpha skeletal muscle; actin, alpha, cardiac 1; actin, alpha, cardiac muscle 1; actin, alpha, vascular smooth muscle; Actin, aortic smooth muscle; Actin, cytoplasmic 2; Actin, cytoplasmic type 5; actin, cytoplasmic, type 5; actin, gamma 1; actin, gamma 1 propeptide; actin, gamma 2, smooth muscle, enteric; actin, gamma, cytoplasmic 1; Actin, gamma-enteric smooth muscle; actin-like protein; Alpha-actin; alpha-actin cardiac; Alpha-actin-1; Alpha-actin-2; Alpha-actin-3; Alpha-cardiac actin; alpha-sma; alpha-smooth muscle actin; alphac-actin; beta-actin FE-3; cardiac actin; Cell growth-inhibiting gene 46 protein; cytoplasmic 2; cytoplasmic beta-actin; cytoplasmic type 5; cytoskeletal gamma-actin; deafness, autosomal dominant 20; deafness, autosomal dominant 26; epididymis luminal protein 176; gamma 1 propeptide; Gamma-2-actin; Gamma-actin; gamma-enteric smooth muscle actin; nemaline myopathy type 3; RP5-1068B5.2; smooth muscle alpha-actin; Smooth muscle gamma-actin; type 5

          View more View less

          Gene Aliases: 0610041G09Rik; a-SMA; AA959943; AAT6; ACT; Act-4; Act4; ACTA; Acta-2; ACTA1; ACTA2; ACTA3; ACTC; Actc-1; ACTC1; ACTE; ACTG; ACTG1; ACTG2; ACTGE; actin; Actl; ACTL3; Acts; ACTSA; ACTSG; Actsk-1; ACTVS; AL023024; alphaSMA; ASD5; ASMA; BRWS2; CFTD; CFTD1; CFTDM; CMD1R; CMH11; cytoplasmic; DFNA20; DFNA26; E51; GAMMA-ACTIN; GIG46; HEL-176; LVNC4; MPFD; MYMY5; NEM1; NEM2; NEM3; RCJMB04_1h13; SHPM; SMalphaA; SMGA; VSCM

          View more View less

          UniProt ID: (Human) P68133, (Rabbit) P62740, (Chicken) P08023, (Human) P62736, (Human) P68032, (Chicken) P68034, (Human) P63261, (Chicken) P53478, (Chicken) P63270, (Human) P63267, (Mouse) P68134, (Rat) P68136, (Rat) P62738, (Mouse) P62737, (Rat) P68035, (Mouse) P68033, (Mouse) P63260, (Rat) P63259, (Mouse) P63268, (Rat) P63269

          View more View less

          Entrez Gene ID: (Chicken) 421534, (Human) 58, (Dog) 488984, (Dog) 477587, (Rabbit) 100009271, (Chicken) 423787, (Human) 59, (Cat) 100144392, (Dog) 478250, (Human) 70, (Chicken) 423298, (Rabbit) 100340339, (Human) 71, (Chicken) 415296, (Dog) 475923, (Chicken) 396084, (Human) 72, (Rabbit) 100350318, (Dog) 475792, (Mouse) 11459, (Rat) 29437, (Rat) 81633, (Mouse) 11475, (Rat) 29275, (Mouse) 11464, (Mouse) 11465, (Rat) 287876, (Mouse) 11468, (Rat) 25365, (Rabbit) 100009506

          View more View less

          Function(s)
          ATP binding structural constituent of cytoskeleton protein binding myosin binding ADP binding protein kinase binding ATPase activity ubiquitin protein ligase binding identical protein binding nucleotide binding molecular_function
          Process(es)
          retina homeostasis movement of cell or subcellular component cell junction assembly Fc-gamma receptor signaling pathway involved in phagocytosis sarcomere organization vascular endothelial growth factor receptor signaling pathway ephrin receptor signaling pathway platelet aggregation skeletal muscle thin filament assembly skeletal muscle fiber development metabolic process actin filament-based movement actomyosin structure organization actin-myosin filament sliding response to drug negative regulation of apoptotic process response to ethanol cardiac myofibril assembly cardiac muscle tissue morphogenesis heart contraction cardiac muscle contraction actin-mediated cell contraction muscle contraction response to mechanical stimulus response to extracellular stimulus response to lithium ion positive regulation of gene expression cell growth muscle filament sliding skeletal muscle fiber adaptation response to steroid hormone mesenchyme migration apoptotic process regulation of blood pressure response to virus vascular smooth muscle contraction glomerular mesangial cell development biological_process response to calcium ion
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0