Aliquoting is unnecessary for -20°C storage.
AgBR1, a mosquito salivary protein also known as A. aegypti bacteria-responsive protein 1, is found to induce inflammatory responses at the bite site (PMID: 32218189). Various studies suggest that AgBR1 plays a role in transmitting or facilitating infections of several viruses via Aedes aegypti mosquitoes (PMID: 30858571, 31312526).