Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Plays a key role in glycolysis and gluconeogenesis. In addition, may also function as scaffolding protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Aldolase 1; aldolase 1, A isoform; Aldolase 2; aldolase 2, B isoform; aldolase A, fructose-bisphosphate; Aldolase B fructose-biphosphate; Aldolase B, fructose-biphosphate; aldolase B, fructose-bisphosphatase; aldolase B, fructose-bisphosphate; epididymis secretory sperm binding protein Li 87p; fructose-1,6-bisphosphate triosephosphate-lyase; Fructose-bisphosphate aldolase A; Fructose-bisphosphate aldolase B; Liver-type aldolase; Lung cancer antigen NY-LU-1; Muscle-type aldolase
Gene Aliases: ALDA; ALDB; Aldo-1; Aldo-2; Aldo1; ALDO2; ALDOA; ALDOB; BC016435; GSD12; HEL-S-87p; LIV10; RNALDOG5
UniProt ID: (Human) P04075, (Human) P05062, (Mouse) P05064, (Rat) P05065, (Mouse) Q91Y97
Entrez Gene ID: (Human) 226, (Human) 229, (Mouse) 11674, (Rat) 24189, (Mouse) 230163, (Rat) 24190
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support