Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 13
Positive Control: U-87 MG cell lysate, RAW264.7 cell lysate, C6 cell lysate, human brain tissue lysate, mouse brain tissue lysate, mouse pancreas tissue lysate, rat brain tissue lysate, rat pancreas tissue lysate, U-87 MG, mouse brain tissue, rat brain tissue.
Tissue Specificity: Group enriched (brain, retina).
Subcellular Location: cytoskeleton, cytosol, extracellular exosome, extracellular region.
Sequence Similarities: 100% Mouse/Rat.
Predicted band size: 110 kDa.
Aldolases are important glycolytic cytosolic enzymes which catalyse the reversible conversion of fructose-1,6-bisphosphate to glyceraldehyde 3-phosphate. There are three aldolase isozymes coded by three distinct genes in mammals, namely aldolases A, B, and C. Aldolase A is heavily expressed in muscle enzyme and aldolase B is heavily expressed in liver and is regarded as a liver specific enzyme. In the adult, aldolase C is the brain-specific isozyme expressed in astrocytes and a few classes of neurons, notably Purkinje cells. Appropriate antibodies to aldolase C are therefore useful to identify astrocytes in cell culture and sectioned material, and the enzyme may be over expressed in some forms of cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ALDA; ALDB; ALDC; Aldolase 1; aldolase 1, A isoform; Aldolase 2; aldolase 2, B isoform; Aldolase 3; aldolase A, fructose-bisphosphate; Aldolase B fructose-biphosphate; Aldolase B, fructose-biphosphate; aldolase B, fructose-bisphosphatase; aldolase B, fructose-bisphosphate; aldolase C, fructose-biphosphate; aldolase C, fructose-bisphosphate; Brain-type aldolase; epididymis secretory sperm binding protein Li 87p; fructoaldolase C; fructose-1,6-biphosphate triosephosphate lyase; fructose-1,6-bisphosphate triosephosphate-lyase; Fructose-bisphosphate aldolase A; Fructose-bisphosphate aldolase B; Fructose-bisphosphate aldolase C; Liver-type aldolase; Lung cancer antigen NY-LU-1; Muscle-type aldolase; Scrapie-responsive protein 2; Zebrin II
Gene Aliases: AI847350; ALDA; ALDB; ALDC; ALDCAA; Aldo-1; Aldo-2; Aldo1; ALDO2; Aldo3; ALDOA; ALDOB; ALDOC; AU040929; BC016435; F16dip7; GSD12; HEL-S-87p; LIV10; RATALDCAA; RNALDOG5; Scrg2
UniProt ID: (Human) P04075, (Human) P05062, (Human) P09972, (Mouse) P05064, (Rat) P05065, (Mouse) Q91Y97, (Mouse) P05063, (Rat) P09117
Entrez Gene ID: (Human) 226, (Human) 229, (Human) 230, (Mouse) 11674, (Rat) 24189, (Mouse) 230163, (Rat) 24190, (Mouse) 11676, (Rat) 24191
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support