Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • ATP1A2 Antibodies

          Alomone Labs, LTD

          Alpha 2 Na+/K+ ATPase Polyclonal Antibody

          View all (6) ATP1A2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Alpha 2 Na+/K+ ATPase Polyclonal Antibody

          • Antibody Testing Data (3)
          Alpha 2 Na+/K+ ATPase Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.
          Alpha 2 Na+/K+ ATPase Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Alpha 2 Na+/K+ ATPase Antibody (ANP-002-200UL) in IHC (F)

          Expression ofNa+/K+ATPase alpha 2 (ATP1A2) in mouse hippocampus and cerebellum - Immunohistochemical staining of perfusion-fixed frozen rat brain sections using Anti-Alpha 2 Na+/K+ATPase Antibody (#ANP-002), (1:300), followed by goat- Anti-rabbit-AlexaFluor-488 secondary Antibody . A. ATP1A2staining (green) in mouse hippocampal CA3 region is labeled in astrocytic processes (arrows surrounding pyramidal cell... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Alpha 2 Na+/K+ ATPase Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Alpha 2 Na+/K+ ATPase Antibody in Western Blot (WB)
          Alpha 2 Na+/K+ ATPase Antibody in Western Blot (WB)

          Product Details

          ANP-002-200UL

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:200
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:300
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          (C)REYSPAATTAENG, corresponding to amino acid residues 7-19 of rat ATP1A2, Intracellular, N-terminus
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          0.8 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 1% BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).

          Target Information

          Atp1a2 encodes an Na+/K+ ATPase, which is a membrane protein that is composed of two subunits - alpha and beta. The Na+/K+ ATPase pump maintains an essential electrochemical gradient within cells through the active transport of sodium (Na+) and potassium (K+) ions. The Atp1a2 encoded protein belongs to the family of P-type cation transport ATPases, and to the subfamily of Na+/K+ -ATPases. Na+/K+ -ATPase is an integral membrane protein responsible for establishing and maintaining the electrochemical gradients of Na and K ions across the plasma membrane. These ion gradients are essential for osmoregulation, for sodium-coupled transport of a variety of organic and inorganic molecules, and for electrical excitability of nerve and muscle. The ATPase enzyme is composed of two subunits, a large catalytic subunit (alpha) and a smaller glycoprotein subunit (beta), and the catalytic subunit of Na+/K+ -ATPase is encoded by multiple genes. The Atp1a2 gene encodes an alpha 2 subunit. Mutations in the Atp1a2 gene result in familial basilar or hemiplegic migraines, and in a rare syndrome known as alternating hemiplegia of childhood. Diseases associated with ATP1A2 include Alternating Hemiplegia Of Childhood and Migraine, Familial Hemiplegic, 2.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ATPase Na+/K+ transporting alpha 2 polypeptide; ATPase, Na+/K+ transporting, alpha 2 polypeptide; ATPase, Na+K+ transporting, alpha 2; Na(+)/K(+) ATPase alpha(+) subunit; Na(+)/K(+) ATPase alpha-2 subunit; Na+/K+ -ATPase alpha 2 subunit; Na+/K+ ATPase, alpha-A(+) catalytic polypeptide; Na+/K+ ATPase, alpha-B polypeptide; Sodium Potassium ATPase; Sodium pump subunit alpha-2; sodium-potassium ATPase catalytic subunit alpha-2; sodium/potassium-transporting ATPase alpha-2 chain; Sodium/potassium-transporting ATPase subunit alpha-2

          View more View less

          Gene Aliases: ATP1A2; Atpa-3; AW060654; FHM2; KIAA0778; MHP2; mKIAA0778; RATATPA2

          View more View less

          UniProt ID: (Human) P50993, (Rat) P06686, (Mouse) Q6PIE5

          View more View less

          Entrez Gene ID: (Human) 477, (Rat) 24212, (Mouse) 98660

          View more View less

          Function(s)
          sodium:potassium-exchanging ATPase activity protein binding ATP binding drug binding ATPase activity potassium ion binding sodium ion binding chaperone binding steroid hormone binding cation-transporting ATPase activity metal ion binding nucleotide binding hydrolase activity
          Process(es)
          neurotransmitter uptake regulation of the force of heart contraction regulation of respiratory gaseous exchange by neurological system process potassium ion transport sodium ion transport cellular sodium ion homeostasis regulation of muscle contraction regulation of smooth muscle contraction regulation of striated muscle contraction metabolic process regulation of blood pressure adult locomotory behavior visual learning potassium ion import regulation of cardiac muscle contraction by calcium ion signaling energy coupled proton transmembrane transport, against electrochemical gradient ATP hydrolysis coupled proton transport regulation of vasoconstriction cellular potassium ion homeostasis response to nicotine sodium ion export from cell locomotion negative regulation of heart contraction negative regulation of striated muscle contraction negative regulation of cytosolic calcium ion concentration relaxation of cardiac muscle cardiac muscle contraction cellular response to mechanical stimulus cellular response to steroid hormone stimulus regulation of cardiac muscle cell contraction negative regulation of calcium ion transmembrane transport negative regulation of calcium:sodium antiporter activity response to glycoside establishment or maintenance of transmembrane electrochemical gradient regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion ion transmembrane transport ATP metabolic process regulation of glutamate uptake involved in transmission of nerve impulse regulation of synaptic transmission, glutamatergic membrane repolarization membrane depolarization during cardiac muscle cell action potential cell communication by electrical coupling involved in cardiac conduction regulation of cardiac conduction potassium ion import across plasma membrane transport ion transport
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0