Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Alpha Sarcoglycan Antibodies

          Invitrogen

          Alpha Sarcoglycan Polyclonal Antibody

          View all (9) Alpha Sarcoglycan antibodies
          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Alpha Sarcoglycan Polyclonal Antibody

          • Antibody Testing Data (4)
          Alpha Sarcoglycan Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Alpha Sarcoglycan Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Alpha Sarcoglycan Antibody (PA5-96273) in ICC/IF

          Immunofluorescence analysis of Alpha Sarcoglycan in L929 cells. Samples were incubated with Alpha Sarcoglycan Polyclonal antibody (Product # PA5-96273) using a dilution of 1:100. Blue: DAPI for nuclear staining. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Alpha Sarcoglycan Antibody in Immunocytochemistry (ICC/IF)
          Alpha Sarcoglycan Antibody in Immunocytochemistry (ICC/IF)
          Alpha Sarcoglycan Antibody in Immunocytochemistry (ICC/IF)
          Alpha Sarcoglycan Antibody in Western Blot (WB)

          Product Details

          PA5-96273

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant fusion protein containing a sequence corresponding to amino acids 60-290 of human alpha Sarcoglycan (alpha Sarcoglycan (SGCA)) (NP_0000141)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.58 mg/mL

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.01% thimerosal

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2808075

          Product Specific Information

          Immunogen sequence: ITYHAHLQGH PDLPRWLRYT QRSPHHPGFL YGSATPEDRG LQVIEVTAYN RDSFDTTRQR LVLEIGDPEG PLLPYQAEFL VRSHDAEEVL PSTPASRFLS ALGGLWEPGE LQLLNVTSAL DRGGRVPLPI EGRKEGVYIK VGSASPFSTC LKMVASPDSH ARCAQGQPPL LSCYDTLAPH FRVDWCNVTL VDKSVPEPAD EVPTPGDGIL EHDPFFCPPT EAPDRDFLVD A; Positive Samples: A-549, LO2, 293T, Mouse skeletal muscle, Mouse heart, Rat skeletal muscle, Rat heart; Cellular Location: Cell membrane, Cytoplasm, Single-pass type I membrane protein, cytoskeleton, sarcolemma

          Target Information

          The sarcoglycan transmembrane proteins are members of the dystrophin complex. Sarcoglycans cluster together to form a complex, which is localized in the cell membrane of skeletal, cardiac, and smooth muscle fibers. Four sarcoglycan subunit proteins, designated alpha-, beta-, gamma- and delta-sarcoglycan, form a complex on the skeletal muscle cell surface membrane. A genetic defect in any one of these proteins causes the loss or marked decrease of the whole sarcoglycan complex, which is observed in the autosomal recessive muscular dystrophy, sarcoglycanopathy. In smooth muscle, beta- and delta-sarcoglycans are associated with epsilon-sarcoglycan, a glycoprotein homologous to alpha-sarcoglycan. Additionally, a complete deficiency in delta-sarcoglycan is the cause of the Syrian hamster BIO.14 cardiomyopathy.

          ⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 50 kDa dystrophin-associated glycoprotein; 50DAG; 50kD DAG; Adhalin; Alpha-sarcoglycan; Alpha-SG; Dystroglycan 2; Dystroglycan-2; Dystroglycan2; limb girdle muscular dystrophy 2D; sarcoglycan, alpha (50kDa dystrophin-associated glycoprotein); SG alpha; SG-alpha

          View more View less

          Gene Aliases: 50DAG; adhalin; ADL; Asg; DAG2; DMDA2; LGMD2D; SCARMD1; SGCA

          View more View less

          UniProt ID: (Human) Q16586, (Mouse) P82350

          View more View less

          Entrez Gene ID: (Human) 6442, (Rat) 303468, (Mouse) 20391

          View more View less

          Function(s)
          calcium ion binding protein binding molecular_function
          Process(es)
          muscle contraction muscle organ development biological_process membrane organization
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-ztd5s:80/100.66.134.6:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0