Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
CASA play an important role in the capacity of milk to transport calcium phosphate. Casoxin D acts as opioid antagonist and has vasorelaxing activity mediated by bradykinin B1 receptors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha S1 casein; Alpha-casein; Alpha-S1-casein; casein alpha-S1; casein, alpha
Gene Aliases: AA959832; AW212882; BOS_6718; CASA; CSN1; CSN1S1; Csna
UniProt ID: (Human) P47710, (Bovine) P02662, (Mouse) P19228
Entrez Gene ID: (Human) 1446, (Bovine) 282208, (Mouse) 12990
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support