Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Angiotensinogen Antibodies

          Bioss

          Angiotensinogen Polyclonal Antibody, Cyanine5.5

          View all (42) Angiotensinogen antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Angiotensinogen Polyclonal Antibody, Cyanine5.5

          Product Details

          BS-20778R-CY55

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:300-5,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:50-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          KLH conjugated synthetic peptide derived from mouse Angiotensinogen
          View immunogen

          Conjugate

          Cyanine5.5 Cyanine5.5 Cyanine5.5

          Excitation/Emission Max

          651/670 nm

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A

          Storage buffer

          TBS, pH 7.4, with 1% BSA, 50% glycerol

          Contains

          0.03% ProClin 300

          Storage conditions

          -20°C or -80°C if preferred

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          The protein encoded by AGT, pre-angiotensinogen or angiotensinogen precursor, is expressed in the liver and is cleaved by the enzyme renin in response to lowered blood pressure. The resulting product, angiotensin I, is then cleaved by angiotensin converting enzyme (ACE) to generate the physiologically active enzyme angiotensin II. The protein is involved in maintaining blood pressure and in the pathogenesis of essential hypertension and preeclampsia. Mutations in this gene are associated with susceptibility to essential hypertension, and can cause renal tubular dysgenesis, a severe disorder of renal tubular development. Defects in this gene have also been associated with non-familial structural atrial fibrillation, and inflammatory bowel disease.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: alpha-1 antiproteinase, antitrypsin; angiotensin I; angiotensin II; angiotensin ll; Angiotensinogen; angiotensinogen (PAT); angiotensinogen (serpin peptidase inhibitor, clade A, member 8); pre-angiotensinogen; serine (or cysteine) proteinase inhibitor; Serpin A8; serpin peptidase inhibitor, clade A, member 8

          View more View less

          Gene Aliases: AGT; AI265500; Ang; AngI; AngII; ANHU; ANRT; Aogen; PAT; SERPINA8

          View more View less

          UniProt ID: (Human) P01019, (Rat) P01015

          View more View less

          Entrez Gene ID: (Human) 183, (Rat) 24179, (Mouse) 11606

          View more View less

          Function(s)
          serine-type endopeptidase inhibitor activity hormone activity protein binding growth factor activity type 1 angiotensin receptor binding type 2 angiotensin receptor binding sodium channel regulator activity angiotensin receptor binding
          Process(es)
          regulation of cell growth positive regulation of cytokine production kidney development blood vessel remodeling angiotensin maturation regulation of blood volume by renin-angiotensin renin-angiotensin regulation of aldosterone production regulation of renal output by angiotensin regulation of heart rate regulation of blood vessel size by renin-angiotensin renal system process angiotensin-mediated drinking behavior positive regulation of extracellular matrix constituent secretion cellular sodium ion homeostasis cell surface receptor signaling pathway G-protein coupled receptor signaling pathway G-protein coupled receptor signaling pathway coupled to cGMP nucleotide second messenger phospholipase C-activating G-protein coupled receptor signaling pathway activation of phospholipase C activity nitric oxide mediated signal transduction cell-cell signaling female pregnancy aging regulation of blood pressure positive regulation of activation of JAK2 kinase activity positive regulation of endothelial cell migration positive regulation of cardiac muscle hypertrophy positive regulation of macrophage derived foam cell differentiation positive regulation of cholesterol esterification negative regulation of endopeptidase activity regulation of norepinephrine secretion positive regulation of phosphatidylinositol 3-kinase signaling artery smooth muscle contraction response to muscle activity involved in regulation of muscle adaptation negative regulation of angiogenesis regulation of vasoconstriction negative regulation of cell growth positive regulation of cellular protein metabolic process positive regulation of superoxide anion generation positive regulation of NAD(P)H oxidase activity negative regulation of tissue remodeling low-density lipoprotein particle remodeling catenin import into nucleus regulation of renal sodium excretion positive regulation of renal sodium excretion regulation of cell proliferation vasodilation positive regulation of nitric oxide biosynthetic process positive regulation of epidermal growth factor receptor signaling pathway positive regulation of blood pressure positive regulation of transcription, DNA-templated positive regulation of vasodilation fibroblast proliferation positive regulation of fibroblast proliferation regulation of long-term neuronal synaptic plasticity smooth muscle cell proliferation cytokine secretion positive regulation of inflammatory response positive regulation of peptidyl-tyrosine phosphorylation positive regulation of NF-kappaB transcription factor activity negative regulation of neurotrophin TRK receptor signaling pathway stress-activated MAPK cascade regulation of calcium ion transport regulation of transmission of nerve impulse cell growth involved in cardiac muscle cell development positive regulation of protein tyrosine kinase activity ERK1 and ERK2 cascade uterine smooth muscle contraction cellular response to mechanical stimulus positive regulation of branching involved in ureteric bud morphogenesis regulation of extracellular matrix assembly positive regulation of gap junction assembly regulation of cardiac conduction positive regulation of reactive oxygen species metabolic process positive regulation of extrinsic apoptotic signaling pathway ovarian follicle rupture blood vessel development branching involved in ureteric bud morphogenesis regulation of systemic arterial blood pressure by circulatory renin-angiotensin angiotensin mediated vasoconstriction involved in regulation of systemic arterial blood pressure renal response to blood flow involved in circulatory renin-angiotensin regulation of systemic arterial blood pressure brain renin-angiotensin system cell-matrix adhesion activation of NF-kappaB-inducing kinase activity excretion establishment of blood-nerve barrier positive regulation of cell proliferation negative regulation of cell proliferation response to cold response to salt stress regulation of gene expression positive regulation of gene expression extracellular matrix organization peristalsis positive regulation of peptidyl-serine phosphorylation positive regulation of multicellular organism growth vasoconstriction hormone metabolic process drinking behavior regulation of apoptotic process positive regulation of catalytic activity positive regulation of MAPK cascade negative regulation of neuron apoptotic process positive regulation of fatty acid biosynthetic process positive regulation of organ growth astrocyte activation regulation of inflammatory response smooth muscle cell differentiation positive regulation of protein kinase C activity positive regulation of membrane hyperpolarization
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0