Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This gene encodes a member of the annexin family, a group of calcium-dependent phospholipid-binding proteins. Annexins have unique N-terminal domains and conserved C-terminal domains, which contain the calcium-dependent phospholipid-binding sites. The encoded protein is a 56-kD antigen recognized by sera from patients with various autoimmune diseases. Transcript variants encoding the same isoform have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 56 kDa autoantigen; Annexin A11; Annexin A11 (Annexin XI) (Calcyclin-associated annexin 50) (CAP-50); Annexin XI; Annexin-11; autoantigen; autoantigen, 56-kD; Calcyclin-associated annexin 50; CAP-50; OTTHUMP00000019957; OTTHUMP00000019958; RP11-369J21
Gene Aliases: A830099O17Rik; ANX11; ANXA11; CAP50
UniProt ID: (Human) P50995, (Rabbit) P33477, (Mouse) P97384
Entrez Gene ID: (Human) 311, (Rabbit) 100009511, (Rat) 290527, (Mouse) 11744
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support