Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • ApoA1 Antibodies
          The PeproTech website has now moved to Thermofisher.com. All orders for PeproTech products must be placed on thermofisher.com or offline via phone, fax, email, or distributor partner. For any questions, please contact PeproTech.CustomerService@thermofisher.com

          Invitrogen

          ApoA1 Polyclonal Antibody, Biotin, PeproTech®

          View all (59) ApoA1 antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite ApoA1 Polyclonal Antibody, Biotin, PeproTech®

          • Antibody Testing Data (3)
          ApoA1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          ApoA1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ApoA1 Antibody (500-P331BT-25UG) in WB

          Western Blot: To detect Human ApoA-I by Western Blot analysis, ApoA1 Polyclonal Antibody, Biotin (Product # 500-P331BT-1MG) can be used at a concentration of 0.1-0.2 µg/mL. When used in conjunction with compatible development reagents, the detection limit for Recombinant Human ApoA-I is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ApoA1 Antibody in Western Blot (WB)
          ApoA1 Antibody in Western Blot (WB)
          ApoA1 Antibody in ELISA (ELISA)

          Product Details

          500-P331BT-25UG

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.1-0.2 µg/mL
          -

          ELISA (ELISA)

          0.25-1.0 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          E.coli-derived, 28.2 kDa Recombinant Human ApoA-I
          View immunogen

          Conjugate

          Biotin Biotin Biotin

          Form

          Lyophilized

          Concentration

          0.1-1.0 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS

          Contains

          no preservative

          Storage conditions

          -20°C

          Shipping conditions

          Ambient

          RRID

          AB_2930466

          Product Specific Information

          AA Sequence of recombinant protein: MDEPPQSPWD RVKDLATVYV DVLKDSGRDY VSQFEGSALG KQLNLKLLDN WDSVTSTFSK LREQLGPVTQ EFWDNLEKET EGLRQEMSKD LEEVKAKVQP YLDDFQKKWQ EEMELYRQKV EPLRAELQEG ARQKLHELQE KLSPLGEEMR DRARAHVDAL RTHLAPYSDE LRQRLAARLE ALKENGGARL AEYHAKATEH LSTLSEKAKP ALEDLRQGLL PVLESFKVSF LSALEEYTKK LNTQ.

          Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human ApoA-I. Anti-Human ApoA-I-specific antibody was purified by affinity chromatography and then biotinylated.

          Sandwich ELISA: To detect Human ApoA-I by sandwich ELISA (using 100 µL/well), a concentration of 0.25-1.0 µg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human ApoA-I (500-P331) as a capture antibody, allows the detection of at least 2000-4000 pg/mL of Recombinant Human ApoA-I.

          Western Blot: To detect Human ApoA-I by Western Blot analysis, this antibody can be used at a concentration of 0.1-0.2 µg/mL. When used in conjunction with compatible development reagents, the detection limit for Recombinant Human ApoA-I is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

          500-P331BT-1MG will be provided as 2 x 500 µg

          Target Information

          Apolipoprotein A-1 (apo A-1) in human plasma is a major protein component of high density lipoprotein (HDL). The molecular mass is 24 kDa. Apo A-1 is a cofactor for enzymes involved in the normal turnover of cholesterol and is produced in the liver. Plasma concentration is 270-380 mg/100 mL.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 1-Sep; 2-Sep; Apo a; apo A-1; APO A1; APO I; Apo-AI; ApoAI; Apolipoprotein; Apolipoprotein A-I; Apolipoprotein A1; Apolipoprotein AI; MGC102525; MGC117399; MGC124929; Udpgt; Ugt1; Ugt1a7

          View more View less

          Gene Aliases: apo(a); APOA1

          View more View less

          UniProt ID: (Human) P02647

          View more View less

          Entrez Gene ID: (Human) 335

          View more View less

          Function(s)
          beta-amyloid binding protein binding phospholipid binding phospholipid transporter activity high-density lipoprotein particle binding cholesterol binding cholesterol transporter activity enzyme binding phosphatidylcholine binding apolipoprotein receptor binding apolipoprotein A-I receptor binding identical protein binding chemorepellent activity lipase inhibitor activity phosphatidylcholine-sterol O-acyltransferase activator activity high-density lipoprotein particle receptor binding
          Process(es)
          retinoid metabolic process regulation of protein phosphorylation endothelial cell proliferation platelet degranulation negative regulation of cytokine secretion involved in immune response phospholipid metabolic process phosphatidylcholine biosynthetic process cholesterol biosynthetic process receptor-mediated endocytosis transforming growth factor beta receptor signaling pathway G-protein coupled receptor signaling pathway integrin-mediated signaling pathway response to nutrient cholesterol metabolic process glucocorticoid metabolic process negative regulation of tumor necrosis factor-mediated signaling pathway positive regulation of cholesterol esterification positive regulation of triglyceride catabolic process negative regulation of very-low-density lipoprotein particle remodeling peripheral nervous system axon regeneration protein oxidation peptidyl-methionine modification triglyceride catabolic process lipid storage regulation of intestinal cholesterol absorption cholesterol transport adrenal gland development organ regeneration neuron projection regeneration regulation of Cdc42 protein signal transduction cholesterol efflux phospholipid efflux negative regulation of heterotypic cell-cell adhesion high-density lipoprotein particle remodeling high-density lipoprotein particle assembly high-density lipoprotein particle clearance positive regulation of Rho protein signal transduction lipoprotein metabolic process lipoprotein biosynthetic process response to drug cholesterol homeostasis blood vessel endothelial cell migration response to estrogen reverse cholesterol transport cellular protein metabolic process positive regulation of fatty acid biosynthetic process negative regulation of interleukin-1 beta secretion negative regulation of inflammatory response protein stabilization negative chemotaxis positive regulation of lipoprotein lipase activity vitamin transport positive regulation of hydrolase activity positive regulation of stress fiber assembly transmembrane transport phospholipid homeostasis negative regulation of lipase activity negative regulation of cell adhesion molecule production negative regulation of response to cytokine stimulus triglyceride homeostasis ERK1 and ERK2 cascade cholesterol import positive regulation of substrate adhesion-dependent cell spreading
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5d94444789-k2m5x:80/100.66.128.182:80.
          git-commit: ec3573d8beaf0fd412f2d0ea2dcb8dd14c74098c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0