Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Apolipoprotein A4 Antibodies

          Bioss

          ApoA4 Polyclonal Antibody, HRP

          View all (36) Apolipoprotein A4 antibodies
          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite ApoA4 Polyclonal Antibody, HRP

          Product Details

          BS-2688R-HRP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:300-1:5,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:200-1:400
          -

          ELISA (ELISA)

          1:500-1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          KLH conjugated synthetic peptide derived from human ApoA4
          View immunogen

          Conjugate

          HRP HRP HRP

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A

          Storage buffer

          TBS, pH 7.4, with 1% BSA, 50% glycerol

          Contains

          0.03% ProClin 300

          Storage conditions

          -20°C or -80°C if preferred

          Shipping conditions

          Wet ice

          Target Information

          Apolipoprotein A4 (also known as ApoA-IV) is a plasma protein that is an O-linked glycoprotein after proteolytic processing. ApoA4 is a major constituent of HDL and it also can be found on the surface of newly synthesized chylomicrons. The physiological function of ApoA4 is not completely identified so far, but evidence from several in vitro studies indicate that this apolipoprotein is involved in several steps of the reverse cholesterol transport pathway, which transports cholesterol from peripheral cells back to the liver.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Apo-AIV; apoA-IV; Apolipoprotein A-IV; Apolipoprotein A4; MGC142154; MGC142156

          View more View less

          Gene Aliases: Apo-AIV; Apoa-4; ApoA-IV; APOA4; apoAIV

          View more View less

          UniProt ID: (Human) P06727, (Mouse) P06728, (Rat) P02651

          View more View less

          Entrez Gene ID: (Human) 337, (Mouse) 11808, (Rat) 25080

          View more View less

          Function(s)
          lipid transporter activity copper ion binding protein binding lipid binding cholesterol binding antioxidant activity cholesterol transporter activity phosphatidylcholine binding protein homodimerization activity phosphatidylcholine-sterol O-acyltransferase activator activity
          Process(es)
          innate immune response in mucosa cholesterol biosynthetic process transport lipid transport response to lipid hydroperoxide leukocyte cell-cell adhesion cholesterol metabolic process positive regulation of cholesterol esterification positive regulation of triglyceride catabolic process removal of superoxide radicals triglyceride catabolic process regulation of intestinal cholesterol absorption neuron projection regeneration regulation of cholesterol transport cholesterol efflux phospholipid efflux very-low-density lipoprotein particle remodeling high-density lipoprotein particle assembly negative regulation of plasma lipoprotein particle oxidation lipoprotein metabolic process cholesterol homeostasis hydrogen peroxide catabolic process reverse cholesterol transport multicellular organism lipid catabolic process positive regulation of fatty acid biosynthetic process phosphatidylcholine metabolic process positive regulation of lipoprotein lipase activity lipid homeostasis protein-lipid complex assembly triglyceride homeostasis peripheral nervous system axon regeneration response to food response to triglyceride retinoid metabolic process chylomicron remodeling high-density lipoprotein particle remodeling chylomicron assembly response to stilbenoid lipoprotein biosynthetic process cellular protein metabolic process
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7fd48b9cb8-69xq6:80/100.66.134.130:80.
          git-commit: fe0b5dd8e7a67b57f27a6e2d26c18f8954d91d59
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0