Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen Sequence: EVSADQVATV MWDYFSQLSN NAKEAVEHLQ KSELTQQLNA LFQDKLGEVN TYAGDLQKKL VPFATELHER LAKDSEKLKE EIGKELEELR ARLLPHANEV SQKIGDNLRE LQQRLEPYAD QLRTQVSTQA EQLRRQLTPY AQRMERVLRE NADSLQASLR PHADELKAKI DQNVEELKGR LTPYADEFKV KIDQTVEELR RSLAPYAQDT QEKLNHQLEG LTFQMKKNAE ELKARISASA EELRQRLAPL AEDVRGNLRG NTEGLQKSLA ELGGHLDQQV EEFRRRVEPY GENFNKALVQ QMEQLRQKLG PHAGDVEGHL SFLEKDLRDK VNSFFSTFKE KESQDKTLSL PELEQQQEQQ QEQQQEQVQM LAPLES
Apolipoprotein A4 (also known as ApoA-IV) is a plasma protein that is an O-linked glycoprotein after proteolytic processing. ApoA4 is a major constituent of HDL and it also can be found on the surface of newly synthesized chylomicrons. The physiological function of ApoA4 is not completely identified so far, but evidence from several in vitro studies indicate that this apolipoprotein is involved in several steps of the reverse cholesterol transport pathway, which transports cholesterol from peripheral cells back to the liver.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Apo-AIV; apoA-IV; Apolipoprotein A-IV; Apolipoprotein A4; MGC142154; MGC142156
Gene Aliases: APOA4
UniProt ID: (Human) P06727
Entrez Gene ID: (Human) 337
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support