Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
This antibody has been tested in direct-ELISA
May be an antioxidant enzyme particularly in the developing shoot and photosynthesizing leaf. Involved in the detoxification of alkyl hydroperoxides with reducing equivalents provided through the thioredoxin system.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2-Cys peroxiredoxin BAS1, chloroplastic; 2-Cys Prx A; Thiol-specific antioxidant protein A; Thioredoxin-dependent peroxiredoxin BAS1
Gene Aliases: AT3G11630; BAS1; F24K9.28; T19F11.3
UniProt ID: (Arabidopsis) Q96291
Entrez Gene ID: (Arabidopsis) 820335
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support