Search Thermo Fisher Scientific
FIGURE: 1 / 2
Positive Control: Rice tissue lysates, Arabidopsis thaliana tissue lysates.
Subcellular Location: Chloroplast, Plastid.
Converts the prephenate produced from the shikimate-chorismate pathway into phenylalanine.