Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: PSETHILTPA SEPVFYQVME RVQQAVWEHQ RTLSPVPLQL DRLGNIWRPW LQPCCGPFPL CWCLREDDPQ; Positive Samples: Mouse liver, Rat liver; Cellular Location: Golgi apparatus, Golgi stack
Arylsulfatase E is a member of the sulfatase family. It is glycosylated postranslationally and localized to the golgi apparatus. Sulfatases are essential for the correct composition of bone and cartilage matrix. X-linked chondrodysplasia punctata, a disease characterized by abnormalities in cartilage and bone development, has been linked to mutations in this gene.