Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Asialo GM1 Antibodies

          Invitrogen

          Asialo GM1 Polyclonal Antibody, Functional Grade, eBioscience™

          7 References
          View all (2) Asialo GM1 antibodies
          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Asialo GM1 Polyclonal Antibody, Functional Grade, eBioscience™

          • Antibody Testing Data (1)
          Asialo GM1 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          Asialo GM1 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Asialo GM1 Antibody (16-6507-39) in Flow

          Staining of C57Bl/6 splenocytes with Anti-Mouse NK1-1 PerCP-Cy5-5 (Product # 45-5941-82) (left) or Anti-Mouse CD11b APC (Product # 17-0112-82) (right) and Anti-Asialo-GM1 Functional Grade Purified.An Anti-Rabbit PE (Product # 12-4739-81) was used for detection.Total viable cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Asialo GM1 Antibody in Flow Cytometry (Flow)

          Product Details

          16-6507-39

          Applications
          Tested Dilution
          Publications

          Immunocytochemistry (ICC/IF)

          Assay-Dependent
          -

          Flow Cytometry (Flow)

          Assay-Dependent
          View 1 publication 1 publication

          Neutralization (Neu)

          -
          View 2 publications 2 publications

          Functional Assay (Functional)

          Assay-Dependent
          View 2 publications 2 publications

          In vitro Assay (IV)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (MISC)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Bovine, Hamster, Human, Mouse, Rat

          Published species

          Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Conjugate

          Functional Grade Functional Grade Functional Grade

          Form

          Lyophilized

          Purification

          Ammonium sulfate precipitation

          Storage buffer

          PBS

          Contains

          no preservative

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_10718540

          Product Specific Information

          Description: This rabbit polyclonal antibody reacts with the asialo ganglio-N-tetraosylceramide (asialo-GM1), a glycolipid on human, mouse, rat, bovine and hamster cells. Glycolipids such as this sialyated version are defined by the specific transferases that modify the lipid. Glycolipids are targets of immune cell response. Auto-antibodies have also been identified. Additionally these glycolipids can act as co-receptors for pathogens. Asialo-GM1 is present on both NK cells and a subset of monocyte/macrophages from the mouse spleen. Expression is also found on rat and bovine brain, fetal liver and on fetal immature thymocytes. Expression has also been shown to increase with tumor progression. The antibody recognizes asialo-GM1 specifically and not other glycolipids, including GM1 and asialo-GM2.

          This antibody has been reported to deplete NK cells in mice and hamsters in vivo and exhibits in vitro effects in a variety of species.

          This polyclonal antibody has been reported for use in flow cytometric analysis, depletion of NK cells, and in vitro functional studies. Intravenous and IP injection of 20-50 µL has been shown to decrease NK1.1 populations after 3-4 days. Repeat injections may be necessary.

          Applications Tested: This polyclonal antibody against asialo-GM1 has been tested by flow cytometric analysis of mouse splenocytes. Optimal staining concentration will vary depending on the lot. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Endotoxin: Less than 0.001 ng/µg antibody as determined by the LAL assay.

          Storage and handling: It is recommended to add 1 mL of sterile (endotoxin free) room temperature water. After reconstitution pass through 0.22 µm filter prior to injection.

          Target Information

          Asialo ganglio-N-tetraosylceramide (asialo-GM1) is a glycolipid on human, mouse, rat, bovine and hamster cells. Glycolipids such as this sialyated version are defined by the specific transferases that modify the lipid. Glycolipids are targets of immune cell response. Auto-antibodies have also been identified. Additionally these glycolipids can act as co-receptors for pathogens. Asialo-GM1 is present on both NK cells and a subset of monocyte/macrophages from the mouse spleen. Expression is also found on rat and bovine brain, fetal liver and on fetal immature thymocytes. Expression has also been shown to increase with tumor progression.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7fd48b9cb8-69xq6:80/100.66.134.130:80.
          git-commit: fe0b5dd8e7a67b57f27a6e2d26c18f8954d91d59
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0