Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • AXIN1 Antibodies

          Bioss

          Axin1 Polyclonal Antibody

          View all (26) AXIN1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Axin1 Polyclonal Antibody

          Product Details

          BS-2439R

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:300
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          Assay-dependent
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:100-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -

          ELISA (ELISA)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          KLH conjugated synthetic peptide derived from human Axin1, amino acids 761-862.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A

          Storage buffer

          0.01M TBS, pH 7.4, with 1% BSA, 50% glycerol

          Contains

          0.02% ProClin 300

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          AXIN1 is a cytoplasmic protein which contains a regulation of G-protein signaling (RGS) domain and a disheveled and axin (DIX) domain and is thought to function as a negative regulator of the WNT signaling pathway that regulates embryonic axis formation. AXIN1 interacts with adenomatosis polyposis coli (APC), beta-catenin, glycogen synthase kinase 3 beta, forming a tetrameric complex resulting in the regulation of the stabilization of beta-catenin. Mutations in the AXIN1 gene have been associated various carcinomas, indicating that it also functions as a tumor suppressor.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Axin-1; Axis inhibition protein 1; axis inhibitor 1; fused, mouse, homolog of; GSK-3beta interacting protein rAxin; hAxin; LA16c-314G4.3; MGC52315; Protein Fused; protein phosphatase 1, regulatory subunit 49; rAxin

          View more View less

          Gene Aliases: AI316800; AXIN; AXIN1; Fu; fused; Kb; Ki; kinky; knobbly; PPP1R49

          View more View less

          UniProt ID: (Human) O15169, (Rat) O70239, (Mouse) O35625

          View more View less

          Entrez Gene ID: (Human) 8312, (Rat) 79257, (Mouse) 12005

          View more View less

          Function(s)
          signal transducer activity GTPase activator activity protein binding beta-catenin binding enzyme binding protein kinase binding receptor signaling complex scaffold activity ubiquitin protein ligase binding protein complex scaffold identical protein binding protein homodimerization activity SMAD binding armadillo repeat domain binding I-SMAD binding p53 binding receptor binding protein C-terminus binding protein domain specific binding protein self-association R-SMAD binding
          Process(es)
          protein polyubiquitination in utero embryonic development optic placode formation regulation of protein phosphorylation positive regulation of protein phosphorylation nucleocytoplasmic transport apoptotic process activation of JUN kinase activity determination of left/right symmetry sensory perception of sound adult walking behavior cell death dorsal/ventral axis specification dorsal/ventral pattern formation Wnt signaling pathway hemoglobin metabolic process forebrain anterior/posterior pattern specification Wnt-activated signaling pathway involved in forebrain neuron fate commitment protein catabolic process negative regulation of Wnt signaling pathway positive regulation of transforming growth factor beta receptor signaling pathway olfactory placode formation cytoplasmic microtubule organization positive regulation of protein ubiquitination activation of protein kinase activity positive regulation of proteasomal ubiquitin-dependent protein catabolic process erythrocyte homeostasis negative regulation of transcription elongation from RNA polymerase II promoter regulation of catenin import into nucleus post-anal tail morphogenesis termination of G-protein coupled receptor signaling pathway positive regulation of JUN kinase activity positive regulation of GTPase activity cellular protein complex assembly negative regulation of fat cell differentiation positive regulation of protein catabolic process positive regulation of protein kinase activity positive regulation of transcription, DNA-templated positive regulation of JNK cascade embryonic eye morphogenesis axial mesoderm development axial mesoderm formation neurological system process negative regulation of protein metabolic process protein homooligomerization positive regulation of ubiquitin-protein transferase activity muscle cell development canonical Wnt signaling pathway embryonic skeletal joint morphogenesis canonical Wnt signaling pathway involved in neural plate anterior/posterior pattern formation cellular response to organic cyclic compound genetic imprinting negative regulation of canonical Wnt signaling pathway Wnt signaling pathway involved in somitogenesis positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process signal transduction multicellular organism development cell differentiation positive regulation of peptidyl-threonine phosphorylation positive regulation of peptidyl-serine phosphorylation proteasome-mediated ubiquitin-dependent protein catabolic process positive regulation of canonical Wnt signaling pathway beta-catenin destruction complex assembly beta-catenin destruction complex disassembly
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-bff4f7d6d-tmrcx:80/100.66.135.11:80.
          git-commit: 683fd26f943b2dde6a08880618459b58d20c3896
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-Offline