Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
Adding 0.2 mL of distilled water will yield a concentration of 500 µg/mL.
Positive Control - WB: monkey COS-7 whole cell, human Jurkat whole cell, rat C6 whole cell, mouse testis tissue. ICC/IF: MCF-7 cell. Flow: HepG2 cell.|Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
BAZ1A may play a role in transcriptional regulation. May be involved in the formation or maintenance of heterochromatin playing a critical role in developmental control.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-dependent chromatin remodeling protein; ATP-dependent chromatin-remodeling protein; ATP-utilizing chromatin assembly and remodeling factor 1; Bromodomain adjacent to zinc finger domain protein 1A; bromodomain adjacent to zinc finger domain, 1A; CHRAC subunit ACF1; gene trap locus 5; hACF1; hWALp1; WCRF180; Williams syndrome transcription factor-related chromatin-remodeling factor 180
Gene Aliases: ACF1; B930060C03; BAZ1A; BC065123; cbp146; Gtl5; hACF1; HSPC317; WALp1; WCRF180
UniProt ID: (Human) Q9NRL2
Entrez Gene ID: (Human) 11177, (Rat) 314126, (Mouse) 217578
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support