Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • BAZ2A Antibodies

          Abnova

          BAZ2A Polyclonal Antibody, MaxPab™

          View all (6) BAZ2A antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite BAZ2A Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (1)
          BAZ2A Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          BAZ2A Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          BAZ2A Antibody (H00011176-B01P) in WB

          Western Blot analysis of BAZ2A expression in transfected 293T cell line by BAZ2A MaxPab polyclonal antibody. Lane 1: BAZ2A transfected lysate(209.55 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          BAZ2A Antibody in Western Blot (WB)

          Product Details

          H00011176-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          BAZ2A (NP_038477.2, 1 a.a. approximately 1905 a.a) full-length human protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MEMEANDHFN FTGLPPAPAA SGLKPSPSSG EGLYTNGSPM NFPQQGKSLN GDVNVNGLST VSHTTTSGIL NSAPHSSSTS HLHHPSVAYD CLWNYSQYPS ANPGSNLKDP PLLSQFSGGQ YPLNGILGGS RQPSSPSHNT NLRAGSQEFW ANGTQSPMGL NFDSQELYDS FPDQNFEVMP NGPPSFFTSP QTSPMLGSSI QTFAPSQEVG SGIHPDEAAE KEMTSVVAEN GTGLVGSLEL EEEQPELKMC GYNGSVPSVE SLHQEVSVLV PDPTVSCLDD PSHLPDQLED TPILSEDSLE PFNSLAPEPV SGGLYGIDDT ELMGAEDKLP LEDSPVISAL DCPSLNNATA FSLLADDSQT STSIFASPTS PPVLGESVLQ DNSFDLNNGS DAEQEEMETQ SSDFPPSLTQ PAPDQSSTIQ LHPATSPAVS PTTSPAVSLV VSPAASPEIS PEVCPAASTV VSPAVFSVVS PASSAVLPAV SLEVPLTASV TSPKASPVTS PAAAFPTASP ANKDVSSFLE TTADVEEITG EGLTASGSGD VMRRRIATPE EVRLPLQHGW RREVRIKKGS HRWQGETWYY GPCGKRMKQF PEVIKYLSRN VVHSVRREHF SFSPRMPVGD FFEERDTPEG LQWVQLSAEE IPSRIQAITG KRGRPRNTEK AKTKEVPKVK RGRGRPPKVK ITELLNKTDN RPLKKLEAQE TLNEEDKAKI AKSKKKMRQK VQRGECQTTI QGQARNKRKQ ETKSLKQKEA KKKSKAEKEK GKTKQEKLKE KVKREKKEKV KMKEKEEVTK AKPACKADKT LATQRRLEER QRQQMILEEM KKPTEDMCLT DHQPLPDFSR VPGLTLPSGA FSDCLTIVEF LHSFGKVLGF DPAKDVPSLG VLQEGLLCQG DSLGEVQDLL VRLLKAALHD PGFPSYCQSL KILGEKVSEI PLTRDNVSEI LRCFLMAYGV EPALCDRLRT QPFQAQPPQQ KAAVLAFLVH ELNGSTLIIN EIDKTLESMS SYRKNKWIVE GRLRRLKTVL AKRTGRSEVE MEGPEECLGR RRSSRIMEET SGMEEEEEEE SIAAVPGRRG RRDGEVDATA SSIPELERQI EKLSKRQLFF RKKLLHSSQM LRAVSLGQDR YRRRYWVLPY LAGIFVEGTE GNLVPEEVIK KETDSLKVAA HASLNPALFS MKMELAGSNT TASSPARARG RPRKTKPGSM QPRHLKSPVR GQDSEQPQAQ LQPEAQLHAP AQPQPQLQLQ LQSHKGFLEQ EGSPLSLGQS QHDLSQSAFL SWLSQTQSHS SLLSSSVLTP DSSPGKLDPA PSQPPEEPEP DEAESSPDPQ ALWFNISAQM PCNAAPTPPP AVSEDQPTPS PQQLASSKPM NRPSAANPCS PVQFSSTPLA GLAPKRRAGD PGEMPQSPTG LGQPKRRGRP PSKFFKQMEQ RYLTQLTAQP VPPEMCSGWW WIRDPEMLDA MLKALHPRGI REKALHKHLN KHRDFLQEVC LRPSADPIFE PRQLPAFQEG IMSWSPKEKT YETDLAVLQW VEELEQRVIM SDLQIRGWTC PSPDSTREDL AYCEHLSDSQ EDITWRGRGR EGLAPQRKTT NPLDLAVMRL AALEQNVERR YLREPLWPTH EVVLEKALLS TPNGAPEGTT TEISYEITPR IRVWRQTLER CRSAAQVCLC LGQLERSIAW EKSVNKVTCL VCRKGDNDEF LLLCDGCDRG CHIYCHRPKM EAVPEGDWFC TVCLAQQVEG EFTQKPGFPK RGQKRKSGYS LNFSEGDGRR RRVLLRGRES PAAGPRYSEE GLSPSKRRRL SMRNHHSDLT FCEIILMEME SHDAAWPFLE PVNPRLVSGY RRIIKNPMDF STMRERLLRG GYTSSEEFAA DALLVFDNCQ TFNEDDSEVG KAGHIMRRFF ESRWEEFYQG KQANL

          Target Information

          TIP 5 is the large subunit of the nucleolar remodeling complex NoRC. NoRC causes the repression of ribosomal gene transcription. It was demonstrated that histone deacetylation is involved in this repression and that TIP5 is associated with the histone deacetylase HDAC1 and mediates the deacetylation of histones in the vicinity of the rDNA promoter. The interaction of TIP5 and HDAC1, which is necessary for transcriptional repression, is established by the C-terminal PHD finger and bromodomain.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Bromodomain adjacent to zinc finger domain protein 2A; bromodomain adjacent to zinc finger domain, 2A; hWALp3; Transcription termination factor I-interacting protein 5; TTF-I interacting peptide 5; TTF-I-interacting protein 5

          View more View less

          Gene Aliases: BAZ2A; KIAA0314; TIP5; WALp3

          View more View less

          UniProt ID: (Human) Q9UIF9

          View more View less

          Entrez Gene ID: (Human) 11176

          View more View less

          Function(s)
          RNA polymerase I CORE element sequence-specific DNA binding RNA binding protein binding zinc ion binding ligand-dependent nuclear receptor binding histone binding lysine-acetylated histone binding
          Process(es)
          chromatin silencing at rDNA DNA methylation chromatin remodeling transcription, DNA-templated regulation of transcription, DNA-templated histone deacetylation
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-8696d955b-mh7vc:80/100.66.135.11:80.
          git-commit: b6496d4449ebd8b46e03b29da6eff4841d0bfc13
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-Offline