Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • BBS5 Antibodies

          Proteintech

          BBS5 Polyclonal Antibody

          33 References
          View all (8) BBS5 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite BBS5 Polyclonal Antibody

          • Antibody Testing Data (17)
          BBS5 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          BBS5 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 17

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          BBS5 Antibody (14569-1-AP) in ICC/IF

          IF result from Dr. Corbit, Kevin. anti-BBS5 (14569-1-AP) marks the ciliary membrane and basal bodies of Human hTERT-RPE1 cells. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          BBS5 Antibody in Immunocytochemistry (ICC/IF)
          BBS5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          BBS5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          BBS5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          BBS5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          BBS5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          BBS5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          BBS5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          BBS5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          BBS5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          BBS5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          BBS5 Antibody in Western Blot (WB)
          BBS5 Antibody in Western Blot (WB)
          BBS5 Antibody in Western Blot (WB)
          BBS5 Antibody in Western Blot (WB)
          BBS5 Antibody in Immunoprecipitation (IP)
          BBS5 Antibody in Immunoprecipitation (IP)

          Product Details

          14569-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:200-1:1,000
          View 13 publications 13 publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:10-1:100
          View 18 publications 18 publications

          Immunoprecipitation (IP)

          0.5-4.0 µg
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          BBS5 Fusion Protein Ag6153 (1-341 aa encoded by BC044593)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.21 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MSVLDALWE DRDVRFDLSA QQMKTRPGEV LIDCLDSIED TKGNNGDRGR LLVTNLRILW HSLALSRVNV SVGYNCILNI TTRTANSKLR GQTEALYILT KCNSTRFEFI FTNLVPGSPR LFTSVMAVHR AYETSKMYRD FKLRSALIQN KQLRLLPQEH VYDKINGVWN LSSDQGNLGT FFITNVRIVW HANMNDSFNV SIPYLQIRSI KIRDSKFGLA LVIESSQQSG GYVLGFKIDP VEKLQESVKE INSLHKVYSA SPIFGVDYEM EEKPQPLEAL TVEQIQDDVE IDSDGHTDAF VAYFADGNKQ QDREPVFSEE LGLAIEKLKD GFTLQGLWEV MS (1-341 aa encoded by BC044593)

          Target Information

          The BBSome complex is thought to function as a coat complex required for sorting of specific membrane proteins to the primary cilia. The BBSome complex is required for ciliogenesis but is dispensable for centriolar satellite function. This ciliogenic function is mediated in part by the Rab8 GDP/GTP exchange factor, which localizes to the basal body and contacts the BBSome. Rab8(GTP) enters the primary cilium and promotes extension of the ciliary membrane. Firstly the BBSome associates with the ciliary membrane and binds to RAB3IP/Rabin8, the guanosyl exchange factor (GEF) for Rab8 and then the Rab8-GTP localizes to the cilium and promotes docking and fusion of carrier vesicles to the base of the ciliary membrane. The BBSome complex, together with the LTZL1, controls SMO ciliary trafficking and contributes to the sonic hedgehog (SHH) pathway regulation. Required for BBSome complex ciliary localization but not for the proper complex assembly.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Bardet-Biedl syndrome 5 homolog; Bardet-Biedl syndrome 5 protein; Bardet-Biedl syndrome 5 protein homolog; BBSome complex member BBS5; novel DUF1448 protein

          View more View less

          Gene Aliases: 1700049I01Rik; 2700023J09Rik; BBS5

          View more View less

          UniProt ID: (Human) Q8N3I7, (Mouse) Q9CZQ9

          View more View less

          Entrez Gene ID: (Human) 129880, (Mouse) 72569, (Rat) 362142

          View more View less

          Function(s)
          RNA polymerase II repressing transcription factor binding protein binding phosphatidylinositol-3-phosphate binding molecular_function
          Process(es)
          transport protein transport cell projection organization cilium assembly intracellular transport heart looping visual perception melanosome transport motile cilium assembly response to stimulus biological_process
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-2w6sl:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0