Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Immunogen Sequence: VAHFTFQPDP EPREYGQTQK MNLFQSVTSA LDNSLAKDPT AVIFGEDVAF GGVFRCTVGL RDKYGKDRVF NTPLCEQGIV GFGIGIAVTG ATAIAEIQFA DYIFPAFDQI VNEAAKYRYR SGDLFNCGSL TIRSPWGCVG HGALYHSQSP EAFFAHCPGI KVVIPRSPFQ AKGLLLSCIE DKNPCIFFEP KILYRAAAEE VPIEPYNIPL SQAEVIQEGS DVTLVAWGTQ VHVIREVASM AKEKLGVSCE VIDLRTIIPW DVDTICKSVI KTGRLLISHE APLTGGFASE ISSTVQEECF LNLEAPISRV CGYDTPFPHI FEPFYIPDKW KCYDALRKMI NY
Branched-chain keto acid dehydrogenase is a multienzyme complex associated with the inner membrane of mitochondria, and functions in the catabolism of branched-chain amino acids. The complex consists of multiple copies of 3 components: branched-chain alpha-keto acid decarboxylase (E1), lipoamide acyltransferase (E2) and lipoamide dehydrogenase (E3). This gene encodes the E1 beta subunit, and mutations therein have been associated with maple syrup urine disease (MSUD), type 1B, a disease characterized by a maple syrup odor to the urine in addition to mental and physical retardation, and feeding problems.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2-oxoisovalerate dehydrogenase beta subunit; 2-oxoisovalerate dehydrogenase subunit beta, mitochondrial; BCKD BCKDH E1-beta; BCKDE1B; BCKDH E1-beta; branched chain alpha-ketoacid dehydrogenase E1-beta subunit; branched chain keto acid dehydrogenase E1 beta; branched chain keto acid dehydrogenase E1, beta polypeptide; Branched-chain alpha-keto acid dehydrogenase E1 component beta chain; dJ279A18.1; E1b-beta subunit of the branched-chain complex; testis secretory sperm-binding protein Li 240mP
Gene Aliases: BCKDE1B; BCKDH E1-beta; BCKDHB; E1B
UniProt ID: (Human) P21953, (Rat) P35738, (Mouse) Q6P3A8
Entrez Gene ID: (Human) 594, (Rat) 29711, (Mouse) 12040
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support