Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MVDPVGFAEA WKAQFPDSEP PRMELRSVGD IEQELERCKA SIRRLEQEVN QERFRMIYLQ TLLAKEKKSY DRQRWGFRRA AQAPDGASEP RASASRPQPA PADGADPPPA EEPEARPDGE GSPGKARPGT ARRPGAAASG ERDDRGPPAS VAALRSNFER IRKGHGQPGA DAEKPFYVNV EFHHERGLVK VNDKEVSDRI SSLGSQAMQM ERKKSQHGAG SSVGDASRPP YRGRSSESSC GVDGDYEDAE LNPRFLKDNL IDANGGSRPP WPPLEYQPYQ SIYVGGMMEG EGKGPLLRSQ STSEQEKRLT WPRRSYSPRS FEDCGGGYTP DCSSNENLTS SEEDFSSGQS SRVSPSPTTY RMFRDKSRSP SQNSQQSFDS SSPPTPQCHK RHRHCPVVVS EATIVGVRKT GQIWPNDGEG AFHGDADGSF GTPPGYGCAA DRAEEQRRHQ DGLPYIDDSP SSSPHLSSKG RGSRDALVSG ALESTKASEL DLEKGLEMRK WVLSGILASE ETYLSHLEAL LLPMKPLKAA ATTSQPVLTS QQIETIFFKV PELYEIHKEF YDGLFPRVQQ WSHQQRVGDL FQKLASQLGV YRAFVDNYGV AMEMAEKCCQ ANAQFAEISE NLRARSNKDA KDPTTKNSLE TLLYKPVDRV TRSTLVLHDL LKHTPASHPD HPLLQDALRI SQNFLSSINE EITPRRQSMT VKKGEHRQLL KDSFMVELVE GARKLRHVFL FTDLLLCTKL KKQSGGKTQQ YDCKWYIPLT DLSFQMVDEL EAVPNIPLVP DEELDALKIK ISQIKSDIQR EKRANKGSKA TERLKKKLSE QESLLLLMSP SMAFRVHSRN GKSYTFLISS DYERAEWREN IREQQKKCFR SFSLTSVELQ MLTNSCVKLQ TVHSIPLTIN KEDDESPGLY GFLNVIVHSA TGFKQSSNLY CTLEVDSFGY FVNKAKTRVY RDTAEPNWNE EFEIELEGSQ TLRILCYEKC YNKTKIPKED GESTDRLMGK GQVQLDPQAL QDRDWQRTVI AMNGIEVKLS VKFNSREFSL KRMPSRKQTG VFGVKIAVVT KRERSKVPYI VRQCVE
A reciprocal translocation between chromosomes 22 and 9 produces the Philadelphia chromosome, which is often found in patients with chronic myelogenous leukemia. The chromosome 22 breakpoint for this translocation is located within the BCR gene. The translocation produces a fusion protein which is encoded by sequence from both BCR and ABL, the gene at the chromosome 9 breakpoint. Although the BCR-ABL fusion protein has been extensively studied, the function of the normal BCR gene product is not clear. The protein has serine/threonine kinase activity and is a GTPase-activating protein for p21rac. Two transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ABL; BCR protein; bcr/abl; BCR/FGFR1 chimera protein; breakpoint cluster region; Breakpoint cluster region protein; c-ABL; c-abl oncogene 1; EC 2.7.11.1; FGFR1/BCR chimera protein; JTK7; p150; Renal carcinoma antigen NY-REN-26; RP11-83J21.1; v-abl
Gene Aliases: ALL; BCR; BCR1; CML; D22S11; D22S662; PHL
UniProt ID: (Human) P11274
Entrez Gene ID: (Human) 613
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support