Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: SLELSAESR MILDAFAQQC SRVLSLLNCG GKLLDSNHSQ SMISCVKQEG SSYNERQEHC HIGKGVHSQT SDNVDIEMQY MQRKQQTSAF LRVFTDSLQN YLLSGSFPTP NPSSASEYGH LADVDPLSTS PVHTLGGWTS PATSESHGHP SSSTLPEEEE EEDEEGYCPR CQELEQEVIS LQQENEELRR KLESIPVPCQ TVLDYLKMVL QHHNQLLIPQ PADQPTEGSK QLLNNYPVYI TSKQWDEAVN SSKKDGRRLL RYLIRFVFTT DELKYSCGL (28-305 aa encoded by BC128592)
BEND4, also named as CCDC4, has 5 isoforms with MW 55-58 kDa, 45-48 kDa and 36 kDa.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BEN domain-containing protein 4; bend4; coiled-coil domain containing 4; Coiled-coil domain-containing protein 4
Gene Aliases: BEND4; CCDC4; D330027G24Rik; EG666938
UniProt ID: (Human) Q6ZU67, (Mouse) P86174
Entrez Gene ID: (Human) 389206, (Mouse) 666938
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support