Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MNIVENSIFL SNLMKSANTF ELKYDLSCEL YRMSTYSTFP AGVPVSERSL ARAGFYYTGV NDKVKCFCCG LMLDNWKRGD SPTEKHKKLY PSCRFVQSLN SVNNLEATSQ PTFPSSVTNS THSLLPGTEN SGYFRGSYSN SPSNPVNSRA NQDFSALMRS SYHCAMNNEN ARLLTFQTWP LTFLSPTDLA KAGFYYIGPG DRVACFACGG KLSNWEPKDN AMSEHLRHFP KCPFIENQLQ DTSRYTVSNL SMQTHAARFK TFFNWPSSVL VNPEQLASAG FYYVGNSDDV KCFCCDGGLR CWESGDDPWV QHAKWFPRCE YLIRIKGQEF IRQVQASYPH LLEQLLSTSD SPGDENAESS IIHFEPGEDH SEDAIMMNTP VINAAVEMGF SRSLVKQTVQ RKILATGENY RLVNDLVLDL LNAEDEIREE ERERATEEKE SNDLLLIRKN RMALFQHLTC VIPILDSLLT AGIINEQEHD VIKQKTQTSL QARELIDTIL VKGNIAATVF RNSLQEAEAV LYEHLFVQQD IKYIPTEDVS DLPVEEQLRR LQEERTCKVC MDKEVSIVFI PCGHLVVCKD CAPSLRKCPI CRSTIKGTVR TFLS
The protein encoded by this gene is a member of a family of proteins that inhibits apoptosis by binding to tumor necrosis factor receptor-associated factors TRAF1 and TRAF2, probably by interfering with activation of ICE-like proteases. The encoded protein inhibits apoptosis induced by serum deprivation but does not affect apoptosis resulting from exposure to menadione, a potent inducer of free radicals. The amino acid sequence predicts three baculovirus IAP repeat domains and a ring finger domain. Transcript variants encoding the same isoform have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AIP 2; API2; Apoptosis inhibitor 2; baculoviral IAP repeat-containing 3; Baculoviral IAP repeat-containing protein 3; C-IAP2; Cellular inhibitor of apoptosis 2; H HIAP1; H MALT2; hIAP-1; IAP homolog; IAP homolog C; IAP-1; Inhibitor of apoptosis protein 1; mammalian IAP homolog C; RING finger protein 49; RING-type E3 ubiquitin transferase BIRC3; TNFR2-TRAF signaling complex protein; TNFR2-TRAF-signaling complex protein 1
Gene Aliases: AIP1; API2; BIRC3; c-IAP2; CIAP2; HAIP1; HIAP1; MALT2; MIHC; RNF49
UniProt ID: (Human) Q13489
Entrez Gene ID: (Human) 330
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support