Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: MAAPQDVHVR ICNQEIVKFD LEVKALIQDI RDCSGPLSAL TELNTKVKEK FQQLRHRIQD LEQLAKEQDK ESEKQLLLQE VENHKKQMLS NQASWRKANL TCKIAIDNLE KAELLQGGDL LRQRKTTKES LAQTSSTITE SLMGISRMMA QQVQQSEEAM QSLVTSSRTI LDANEEFKSM SGTIQLGRKL ITKYNRRELT DKLLIFLALA LFLATVLYIV KKRLFPFL
NIP1 (BNIP1) is a member of the BCL2/adenovirus E1B 19 kd-interacting protein (BNIP) family. It interacts with the E1B 19 kDa protein which is responsible for the protection of virally-induced cell death, as well as E1B 19 kDa-like sequences of BCL2, also an apoptotic protector. Alternative splicing of this gene results in four products of unknown function. Transcript variant BNIP1 contains the entire coding region of the gene. This variant contains a fully conserved BH3 domain, which has been associated with pro-apoptotic function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BCL2/adenovirus E1B 19 kDa protein-interacting protein 1; BCL2/adenovirus E1B 19kDa interacting protein 1; Transformation-related gene 8 protein; TRG-8; Vesicle transport protein SEC20
Gene Aliases: BNIP1; NIP1; SEC20; SEC20L; TRG-8; TRG8
UniProt ID: (Human) Q12981
Entrez Gene ID: (Human) 662
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support