Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: VRSHTGERP FQCCQCSYAS RDTYKLKRHM RTHSGEKPYE CHICHTRFTQ SGTMKIHILQ KHGENVPKYQ CPHCATIIAR KSDLRVHMRN LHAYSAAELK CRYCSAVFHE RYALIQHQKT HKNEKRFKCK HCSYACKQER HMTAHIRTHT GEKPFTCLSC NKCFRQKQLL NAHFRKYHDA NFIPTVYKCS KCGKGFSRWI NLHRHSEKCG SGEAKSAASG KGRRTRKRKQ TILKEATKGQ KEAAKGWKEA ANGDEAAAEE ASTTKGEQFP GEMFPVACRE TTARVKEEVD EGVTCEMLLN TMDK (361-663 aa encoded by B C130486)
CTCFL is a paralog of CTCF and appears to be expressed primarily in the cytoplasm of spermatocytes. CTCFL is normally expressed in a mutually exclusive pattern that correlates with resetting of methylation marks during male germ cell differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BORIS; BORIS like protein; BORIS-like protein; Brother of the regulator of imprinted sites; Cancer/testis antigen 27; CCCTC-binding factor; CCCTC-binding factor (zinc finger protein)-like; CT27; CTCF paralog; CTCF T; CTCF-like protein; HMG 1L1; HMG-1L1; likely orthologue of H. sapiens CCCTC-binding factor-like protein (CTCFL); MGC163358; MGC169105; MGC169106; putative high mobility group protein 1-like 1; putative high mobility group protein B1-like 1; Transcriptional repressor CTCFL; Zinc finger protein CTCF-T
Gene Aliases: BORIS; CT27; CTCF-T; CTCFL; dJ579F20.2; HMGB1L1; OTTMUSG00000016680
UniProt ID: (Human) Q8NI51, (Mouse) A2APF3
Entrez Gene ID: (Human) 140690, (Mouse) 664799
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support