Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MLRILCLALC SLLTGTRADP GALLRLGMDI MNQVQSAMDE SHILEKMAAE AGKKQPGMKP IKGITNLKVK DVQLPVITLN FVPGVGIFQC VSTGMTVTGK SFMGGNMEII VALNITATNR LLRDEETGLP VFKSEGCEVI LVNVKTNLPS NMLPKMVNKF LDSTLHKVLP GLMCPAIDAV LVYVNRKWTN LSDPMPVGQM GTVKYVLMSA PATTASYIQL DFSPVVQQQK GKTIKLADAG EALTFPEGYA KGSSQLLLPA TFLSAELALL QKSFHVNIQD TMIGELPPQT TKTLARFIPE VAVAYPKSKP LTTQIKIKKP PKVTMKTGKS LLHLHSTLEM FAARWRSKAP MSLFLLEVHF NLKVQYSVHE NQLQMATSLD RLLSLSRKSS SIGNFNEREL TGFITSYLEE AYIPVVNDVL QVGLPLPDFL AMNYNLAELD IVENALMLDL KLG
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bactericidal/permeability-increasing protein-like 3; BPI fold containing family B, member 6; BPI fold-containing family B member 6
Gene Aliases: BPIFB6; BPIL3; LPLUNC6
UniProt ID: (Human) Q8NFQ5
Entrez Gene ID: (Human) 128859
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support