Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Bromodomain containing protein 9 (BRD9) is an uncharacterized protein that contains one bromodomain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bromodomain-containing protein 9; DKFZp434D0711; DKFZp686L0539; FLJ43530; Rhabdomyosarcoma antigen MU-RMS-40.8; sarcoma antigen NY-SAR-29
Gene Aliases: AL022779; BRD9; LAVS3040; PRO9856; UNQ3040/PRO9856
UniProt ID: (Human) Q9H8M2, (Mouse) Q3UQU0
Entrez Gene ID: (Human) 65980, (Mouse) 105246
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support