Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: REVISCMFEP EGNACSLTDS TAEEHVLALV EHAADEARDR INRFLPGGKM GYLKRNGDGS LLYSVVNTAE PDADEEETHP VDLSSLSSKL LPGFTTLGFK DERRNKVTFL SSATTALSMQ NNSVFGDLKS DEMELLYSAY GDETGVQCAL SLQEFVKDAG SYSKKVVDDL LDQITGGDHS RTLFQLKQRR NVPMKPPDEA KVGDTLGDSS SSVLEFMSMK SYPDVSVDIS MLSSLGKVKK ELDPDDSHLN LDETTKLLQD LHEAQAERGG SRPSSNLSSL SNASERDQHH LGSPSRLSVG EQPDVTHDPY EFLQSPEPAA SAKT
Bromodomain containing protein 9 (BRD9) is an uncharacterized protein that contains one bromodomain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bromodomain-containing protein 9; DKFZp434D0711; DKFZp686L0539; FLJ43530; Rhabdomyosarcoma antigen MU-RMS-40.8; sarcoma antigen NY-SAR-29
Gene Aliases: AL022779; BRD9; LAVS3040; PRO9856; UNQ3040/PRO9856
UniProt ID: (Human) Q9H8M2, (Mouse) Q3UQU0
Entrez Gene ID: (Human) 65980, (Rat) 308067, (Mouse) 105246
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support